Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 72495337-72495596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6B
Entrez Gene ID
23233
Gene Name
exocyst complex component 6B
Gene Aliases
SEC15B, SEC15L2
UniGene
Hs.303454
Gene Chromosome Location
Chr.2: 72175984-72826042 on Build GRCh38
Transcripts
NM_001321729.1 NM_001321730.1 NM_001321731.1 NM_001321733.1 NM_001321734.1 NM_015189.2 NR_135773.1 NR_135774.1 XM_005264224.1 XM_011532711.2 XM_011532712.2 XM_017003641.1 XM_017003642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5460564 c.124A>T p.I42F Chr.2: 72495470-72495470 on Build GRCh38
1615080 c.63delT p.P22fs*25 Chr.2: 72495531-72495531 on Build GRCh38
1737608 c.1553+3A>G p.? Chr.2: 72495427-72495427 on Build GRCh38
5460563 c.1513A>T p.I505F Chr.2: 72495470-72495470 on Build GRCh38
1737609 c.164+3A>G p.? Chr.2: 72495427-72495427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148711200 Chr.2: 72495464-72495464 on Build GRCh38
rs567034443 Chr.2: 72495519-72495519 on Build GRCh38
rs770640090 Chr.2: 72495500-72495500 on Build GRCh38
rs759323758 Chr.2: 72495524-72495524 on Build GRCh38
rs778726682 Chr.2: 72495442-72495444 on Build GRCh38
rs753247136 Chr.2: 72495421-72495421 on Build GRCh38
rs760464011 Chr.2: 72495498-72495498 on Build GRCh38
rs764910423 Chr.2: 72495401-72495401 on Build GRCh38
rs764814187 Chr.2: 72495531-72495531 on Build GRCh38
rs372560536 Chr.2: 72495460-72495460 on Build GRCh38
rs377015657 Chr.2: 72495536-72495536 on Build GRCh38
rs879004247 Chr.2: 72495507-72495507 on Build GRCh38
rs375972673 Chr.2: 72495463-72495463 on Build GRCh38
rs748047858 Chr.2: 72495468-72495468 on Build GRCh38
rs764610426 Chr.2: 72495549-72495549 on Build GRCh38
rs202201931 Chr.2: 72495436-72495436 on Build GRCh38
rs185661651 Chr.2: 72495396-72495396 on Build GRCh38
rs781416026 Chr.2: 72495444-72495444 on Build GRCh38
rs777965638 Chr.2: 72495434-72495434 on Build GRCh38
rs988438816 Chr.2: 72495366-72495366 on Build GRCh38
rs377739932 Chr.2: 72495534-72495534 on Build GRCh38
rs747355096 Chr.2: 72495435-72495435 on Build GRCh38
rs200787488 Chr.2: 72495380-72495380 on Build GRCh38
rs752019287 Chr.2: 72495566-72495566 on Build GRCh38
rs753448222 Chr.2: 72495395-72495395 on Build GRCh38
rs773048433 Chr.2: 72495497-72495497 on Build GRCh38
rs772008057 Chr.2: 72495469-72495469 on Build GRCh38
rs762580245 Chr.2: 72495542-72495542 on Build GRCh38
rs766010263 Chr.2: 72495390-72495390 on Build GRCh38
rs187503402 Chr.2: 72495448-72495448 on Build GRCh38
rs776422975 Chr.2: 72495518-72495518 on Build GRCh38
rs374318382 Chr.2: 72495465-72495465 on Build GRCh38

View Full Product Details