Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149140130-149140397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS3
Entrez Gene ID
84343
Gene Name
HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
Gene Aliases
BLOC2S1, SUTAL
UniGene
Hs.558314
Gene Chromosome Location
Chr.3: 149129584-149173196 on Build GRCh38
Transcripts
NM_001308258.1 NM_032383.4 XM_005247834.3 XM_017007323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209944 c.402G>T p.E134D Chr.3: 149140188-149140188 on Build GRCh38
1419827 c.530G>A p.R177H Chr.3: 149140316-149140316 on Build GRCh38
1039736 c.399G>A p.S133S Chr.3: 149140185-149140185 on Build GRCh38
1039737 c.559A>G p.T187A Chr.3: 149140345-149140345 on Build GRCh38
7126435 c.526G>C p.E176Q Chr.3: 149140312-149140312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766173769 Chr.3: 149140212-149140212 on Build GRCh38
rs144174562 Chr.3: 149140276-149140276 on Build GRCh38
rs769747723 Chr.3: 149140189-149140189 on Build GRCh38
rs776387542 Chr.3: 149140253-149140253 on Build GRCh38
rs776135012 Chr.3: 149140197-149140197 on Build GRCh38
rs140810728 Chr.3: 149140184-149140184 on Build GRCh38
rs764696921 Chr.3: 149140200-149140200 on Build GRCh38
rs746331416 Chr.3: 149140359-149140359 on Build GRCh38
rs551759358 Chr.3: 149140246-149140246 on Build GRCh38
rs757400160 Chr.3: 149140350-149140350 on Build GRCh38
rs756348740 Chr.3: 149140319-149140319 on Build GRCh38
rs751435609 Chr.3: 149140306-149140306 on Build GRCh38
rs770488643 Chr.3: 149140228-149140230 on Build GRCh38
rs111810124 Chr.3: 149140352-149140352 on Build GRCh38
rs563502898 Chr.3: 149140185-149140185 on Build GRCh38
rs771446773 Chr.3: 149140192-149140192 on Build GRCh38
rs139402303 Chr.3: 149140272-149140272 on Build GRCh38
rs1039060115 Chr.3: 149140327-149140327 on Build GRCh38
rs752455255 Chr.3: 149140210-149140210 on Build GRCh38
rs1036366561 Chr.3: 149140296-149140296 on Build GRCh38
rs774802486 Chr.3: 149140193-149140193 on Build GRCh38
rs752988768 Chr.3: 149140316-149140316 on Build GRCh38
rs995345573 Chr.3: 149140356-149140356 on Build GRCh38
rs368439750 Chr.3: 149140311-149140311 on Build GRCh38
rs758281135 Chr.3: 149140211-149140211 on Build GRCh38
rs762881753 Chr.3: 149140293-149140293 on Build GRCh38
rs756544891 Chr.3: 149140247-149140247 on Build GRCh38
rs772649825 Chr.3: 149140277-149140277 on Build GRCh38
rs779562582 Chr.3: 149140240-149140240 on Build GRCh38
rs375065575 Chr.3: 149140214-149140214 on Build GRCh38
rs779612018 Chr.3: 149140357-149140357 on Build GRCh38
rs750390759 Chr.3: 149140201-149140201 on Build GRCh38
rs777954083 Chr.3: 149140233-149140233 on Build GRCh38
rs766369802 Chr.3: 149140303-149140303 on Build GRCh38
rs200726828 Chr.3: 149140194-149140194 on Build GRCh38
rs773902711 Chr.3: 149140190-149140190 on Build GRCh38
rs780231164 Chr.3: 149140369-149140369 on Build GRCh38
rs559742301 Chr.3: 149140213-149140213 on Build GRCh38
rs755170109 Chr.3: 149140179-149140179 on Build GRCh38
rs979740599 Chr.3: 149140265-149140265 on Build GRCh38
rs368535581 Chr.3: 149140161-149140161 on Build GRCh38
rs369293593 Chr.3: 149140196-149140196 on Build GRCh38
rs866117577 Chr.3: 149140284-149140284 on Build GRCh38
rs772518774 Chr.3: 149140367-149140367 on Build GRCh38
rs754042219 Chr.3: 149140344-149140344 on Build GRCh38
rs748883997 Chr.3: 149140188-149140188 on Build GRCh38
rs765277624 Chr.3: 149140159-149140159 on Build GRCh38
rs767183089 Chr.3: 149140315-149140315 on Build GRCh38
rs778009225 Chr.3: 149140328-149140328 on Build GRCh38
rs756254897 Chr.3: 149140225-149140225 on Build GRCh38
rs752837159 Chr.3: 149140222-149140222 on Build GRCh38
rs146744768 Chr.3: 149140234-149140234 on Build GRCh38
rs529624415 Chr.3: 149140191-149140191 on Build GRCh38
rs577273287 Chr.3: 149140178-149140178 on Build GRCh38
rs758230252 Chr.3: 149140164-149140164 on Build GRCh38
rs368415805 Chr.3: 149140174-149140174 on Build GRCh38
rs772860370 Chr.3: 149140180-149140192 on Build GRCh38

View Full Product Details