Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88558806-88559077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK3
Entrez Gene ID
9451
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
PEK, PERK, WRS
UniGene
Hs.591589
Gene Chromosome Location
Chr.2: 88556740-88627576 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Gene Symbol
LOC101928371
Entrez Gene ID
101928371
Gene Name
uncharacterized LOC101928371
Gene Aliases
-
UniGene
Hs.738045
Gene Chromosome Location
Chr.2: 88538720-88575610 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3840219 c.3150G>T p.E1050D Chr.2: 88558917-88558917 on Build GRCh38
5344276 c.3122C>T p.P1041L Chr.2: 88558945-88558945 on Build GRCh38
1637915 c.3100G>A p.V1034I Chr.2: 88558967-88558967 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776519954 Chr.2: 88559026-88559026 on Build GRCh38
rs373029207 Chr.2: 88559005-88559005 on Build GRCh38
rs995615422 Chr.2: 88558970-88558970 on Build GRCh38
rs907121032 Chr.2: 88558863-88558863 on Build GRCh38
rs747716914 Chr.2: 88558983-88558983 on Build GRCh38
rs762125383 Chr.2: 88558916-88558916 on Build GRCh38
rs776609488 Chr.2: 88558873-88558873 on Build GRCh38
rs756319874 Chr.2: 88558956-88558956 on Build GRCh38
rs772535408 Chr.2: 88559010-88559014 on Build GRCh38
rs143199364 Chr.2: 88558849-88558849 on Build GRCh38
rs751710229 Chr.2: 88558886-88558886 on Build GRCh38
rs576434875 Chr.2: 88558862-88558862 on Build GRCh38
rs543627465 Chr.2: 88558878-88558878 on Build GRCh38
rs770494712 Chr.2: 88559022-88559022 on Build GRCh38
rs112399417 Chr.2: 88558903-88558903 on Build GRCh38
rs759995461 Chr.2: 88559005-88559010 on Build GRCh38
rs574983475 Chr.2: 88559013-88559013 on Build GRCh38
rs938556820 Chr.2: 88558892-88558892 on Build GRCh38
rs538943881 Chr.2: 88558991-88559003 on Build GRCh38
rs1055612743 Chr.2: 88558898-88558898 on Build GRCh38
rs762127998 Chr.2: 88558833-88558833 on Build GRCh38
rs780698374 Chr.2: 88558965-88558965 on Build GRCh38
rs767840632 Chr.2: 88558928-88558928 on Build GRCh38
rs777469262 Chr.2: 88558988-88558988 on Build GRCh38
rs769528079 Chr.2: 88558980-88558981 on Build GRCh38
rs759519501 Chr.2: 88559029-88559029 on Build GRCh38
rs545376650 Chr.2: 88558868-88558868 on Build GRCh38
rs146129461 Chr.2: 88558963-88558963 on Build GRCh38
rs770872586 Chr.2: 88558869-88558869 on Build GRCh38
rs945691985 Chr.2: 88558854-88558854 on Build GRCh38
rs767428404 Chr.2: 88558938-88558938 on Build GRCh38
rs747945365 Chr.2: 88558900-88558901 on Build GRCh38
rs750529763 Chr.2: 88558953-88558953 on Build GRCh38
rs561904423 Chr.2: 88558894-88558894 on Build GRCh38
rs771764830 Chr.2: 88558994-88558994 on Build GRCh38
rs779355571 Chr.2: 88558977-88558977 on Build GRCh38
rs374423016 Chr.2: 88558967-88558967 on Build GRCh38
rs759322909 Chr.2: 88558876-88558876 on Build GRCh38
rs76949751 Chr.2: 88558848-88558848 on Build GRCh38
rs770369906 Chr.2: 88558988-88558991 on Build GRCh38
rs748961764 Chr.2: 88558988-88558990 on Build GRCh38
rs755369984 Chr.2: 88558972-88558972 on Build GRCh38

View Full Product Details