Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92468792-92469065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837455 c.2294G>A p.R765Q Chr.10: 92468905-92468905 on Build GRCh38
1210230 c.2253A>C p.E751D Chr.10: 92468946-92468946 on Build GRCh38
346279 c.2320+1G>A p.? Chr.10: 92468878-92468878 on Build GRCh38
7180709 c.2302G>C p.E768Q Chr.10: 92468897-92468897 on Build GRCh38
3441530 c.2279C>T p.P760L Chr.10: 92468920-92468920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763366654 Chr.10: 92468880-92468880 on Build GRCh38
rs753657552 Chr.10: 92468956-92468956 on Build GRCh38
rs768368186 Chr.10: 92468869-92468869 on Build GRCh38
rs374103212 Chr.10: 92469013-92469013 on Build GRCh38
rs1019582649 Chr.10: 92468939-92468939 on Build GRCh38
rs781447234 Chr.10: 92468907-92468907 on Build GRCh38
rs377323965 Chr.10: 92468863-92468863 on Build GRCh38
rs112372000 Chr.10: 92468990-92468990 on Build GRCh38
rs771585390 Chr.10: 92468883-92468883 on Build GRCh38
rs752808781 Chr.10: 92468931-92468931 on Build GRCh38
rs746094977 Chr.10: 92468935-92468935 on Build GRCh38
rs760053435 Chr.10: 92468906-92468906 on Build GRCh38
rs765871360 Chr.10: 92469004-92469004 on Build GRCh38
rs771523149 Chr.10: 92468841-92468841 on Build GRCh38
rs546860255 Chr.10: 92468826-92468826 on Build GRCh38
rs764233258 Chr.10: 92468952-92468952 on Build GRCh38
rs760898950 Chr.10: 92468944-92468944 on Build GRCh38
rs755627829 Chr.10: 92468894-92468894 on Build GRCh38
rs773719072 Chr.10: 92468873-92468873 on Build GRCh38
rs200555551 Chr.10: 92468987-92468987 on Build GRCh38
rs373755533 Chr.10: 92468904-92468904 on Build GRCh38
rs114285269 Chr.10: 92468974-92468974 on Build GRCh38
rs1017085897 Chr.10: 92468857-92468857 on Build GRCh38
rs374934906 Chr.10: 92468831-92468831 on Build GRCh38
rs767559427 Chr.10: 92468929-92468929 on Build GRCh38
rs777570320 Chr.10: 92468829-92468829 on Build GRCh38
rs367995610 Chr.10: 92469029-92469029 on Build GRCh38
rs771152795 Chr.10: 92468905-92468905 on Build GRCh38
rs779614549 Chr.10: 92468847-92468847 on Build GRCh38
rs757095498 Chr.10: 92468968-92468968 on Build GRCh38
rs774286254 Chr.10: 92468961-92468961 on Build GRCh38
rs962858866 Chr.10: 92468879-92468879 on Build GRCh38
rs746463293 Chr.10: 92469011-92469011 on Build GRCh38
rs778775368 Chr.10: 92468833-92468833 on Build GRCh38
rs745796705 Chr.10: 92468836-92468836 on Build GRCh38
rs771458777 Chr.10: 92469036-92469036 on Build GRCh38

View Full Product Details