Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37443935-37444164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD2
Entrez Gene ID
57495
Gene Name
NACHT and WD repeat domain containing 2
Gene Aliases
KIAA1239
UniGene
Hs.4280
Gene Chromosome Location
Chr.4: 37245068-37449465 on Build GRCh38
Transcripts
NM_001144990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161939 c.2037T>A p.D679E Chr.4: 37444025-37444025 on Build GRCh38
1429516 c.2136delG p.E713fs*7 Chr.4: 37444124-37444124 on Build GRCh38
1429515 c.2094C>A p.S698R Chr.4: 37444082-37444082 on Build GRCh38
5197923 c.2004_2005insA p.M670fs*5 Chr.4: 37443992-37443993 on Build GRCh38
5446776 c.1992C>T p.Y664Y Chr.4: 37443980-37443980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370627496 Chr.4: 37444002-37444002 on Build GRCh38
rs745573547 Chr.4: 37444110-37444110 on Build GRCh38
rs897631614 Chr.4: 37444044-37444044 on Build GRCh38
rs78505176 Chr.4: 37443965-37443965 on Build GRCh38
rs865867421 Chr.4: 37443985-37443985 on Build GRCh38
rs1043772236 Chr.4: 37443989-37443989 on Build GRCh38
rs751000613 Chr.4: 37444054-37444054 on Build GRCh38
rs892376170 Chr.4: 37443964-37443964 on Build GRCh38
rs1040212999 Chr.4: 37444006-37444006 on Build GRCh38
rs867747769 Chr.4: 37444072-37444072 on Build GRCh38
rs887469979 Chr.4: 37444102-37444102 on Build GRCh38
rs570435418 Chr.4: 37444127-37444127 on Build GRCh38
rs540187242 Chr.4: 37444076-37444076 on Build GRCh38
rs181518983 Chr.4: 37444129-37444129 on Build GRCh38
rs777717356 Chr.4: 37444005-37444005 on Build GRCh38
rs141630665 Chr.4: 37444105-37444105 on Build GRCh38
rs993430911 Chr.4: 37444058-37444058 on Build GRCh38
rs1027553692 Chr.4: 37444082-37444082 on Build GRCh38
rs904967913 Chr.4: 37443995-37443995 on Build GRCh38
rs756727856 Chr.4: 37444055-37444055 on Build GRCh38

View Full Product Details