Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197347207-197347476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRB1
Entrez Gene ID
23418
Gene Name
crumbs 1, cell polarity complex component
Gene Aliases
LCA8, RP12
UniGene
Hs.126135
Gene Chromosome Location
Chr.1: 197201462-197478455 on Build GRCh38
Transcripts
NM_001193640.1 NM_001257965.1 NM_001257966.1 NM_201253.2 NR_047563.1 NR_047564.1 XM_011509365.2 XM_011509367.1 XM_017000851.1 XM_017000852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6286394 c.947G>A p.C316Y Chr.1: 197347438-197347438 on Build GRCh38
5969044 c.762C>T p.Y254Y Chr.1: 197347460-197347460 on Build GRCh38
115932 c.866C>T p.T289M Chr.1: 197347357-197347357 on Build GRCh38
5379060 c.678G>A p.G226G Chr.1: 197347376-197347376 on Build GRCh38
5941490 c.849-7T>G p.? Chr.1: 197347333-197347333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757239609 Chr.1: 197347377-197347377 on Build GRCh38
rs371751359 Chr.1: 197347360-197347360 on Build GRCh38
rs1005514724 Chr.1: 197347275-197347275 on Build GRCh38
rs780792667 Chr.1: 197347380-197347380 on Build GRCh38
rs758266571 Chr.1: 197347301-197347301 on Build GRCh38
rs375065689 Chr.1: 197347317-197347317 on Build GRCh38
rs779835125 Chr.1: 197347420-197347420 on Build GRCh38
rs368747600 Chr.1: 197347300-197347300 on Build GRCh38
rs913193664 Chr.1: 197347235-197347235 on Build GRCh38
rs375379774 Chr.1: 197347359-197347359 on Build GRCh38
rs1337167 Chr.1: 197347305-197347305 on Build GRCh38
rs994294338 Chr.1: 197347331-197347331 on Build GRCh38
rs888455609 Chr.1: 197347261-197347261 on Build GRCh38
rs779099511 Chr.1: 197347368-197347368 on Build GRCh38
rs147244321 Chr.1: 197347358-197347358 on Build GRCh38
rs745662323 Chr.1: 197347381-197347381 on Build GRCh38
rs768083920 Chr.1: 197347249-197347249 on Build GRCh38
rs569735105 Chr.1: 197347244-197347244 on Build GRCh38
rs773946688 Chr.1: 197347431-197347431 on Build GRCh38
rs1572514 Chr.1: 197347278-197347278 on Build GRCh38
rs575263031 Chr.1: 197347332-197347332 on Build GRCh38
rs765228022 Chr.1: 197347448-197347448 on Build GRCh38
rs542716912 Chr.1: 197347376-197347376 on Build GRCh38
rs911156007 Chr.1: 197347405-197347405 on Build GRCh38
rs62636263 Chr.1: 197347357-197347357 on Build GRCh38
rs996444190 Chr.1: 197347379-197347379 on Build GRCh38
rs368366574 Chr.1: 197347344-197347344 on Build GRCh38
rs748995167 Chr.1: 197347424-197347424 on Build GRCh38
rs138168156 Chr.1: 197347349-197347349 on Build GRCh38
rs189235074 Chr.1: 197347243-197347243 on Build GRCh38
rs772387626 Chr.1: 197347299-197347299 on Build GRCh38
rs865828487 Chr.1: 197347417-197347417 on Build GRCh38
rs1046799192 Chr.1: 197347343-197347343 on Build GRCh38
rs554706901 Chr.1: 197347400-197347400 on Build GRCh38
rs756362313 Chr.1: 197347296-197347296 on Build GRCh38
rs775070124 Chr.1: 197347334-197347334 on Build GRCh38
rs988534225 Chr.1: 197347338-197347338 on Build GRCh38
rs988928414 Chr.1: 197347234-197347234 on Build GRCh38
rs901279546 Chr.1: 197347240-197347240 on Build GRCh38
rs777566954 Chr.1: 197347303-197347303 on Build GRCh38
rs768247556 Chr.1: 197347425-197347425 on Build GRCh38
rs978443271 Chr.1: 197347384-197347384 on Build GRCh38
rs148240351 Chr.1: 197347354-197347354 on Build GRCh38
rs539347284 Chr.1: 197347304-197347304 on Build GRCh38
rs747523008 Chr.1: 197347330-197347330 on Build GRCh38

View Full Product Details