Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100058555-100058808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14A
Entrez Gene ID
64645
Gene Name
major facilitator superfamily domain containing 14A
Gene Aliases
HIAT1
UniGene
Hs.124156
Gene Chromosome Location
Chr.1: 100038078-100083377 on Build GRCh38
Transcripts
NM_033055.2 XM_005271118.1 XM_011541957.1 XM_017002084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412795 c.224G>A p.G75D Chr.1: 100058709-100058709 on Build GRCh38
5525052 c.232C>T p.Q78* Chr.1: 100058717-100058717 on Build GRCh38
6679788 c.199C>T p.P67S Chr.1: 100058684-100058684 on Build GRCh38
5562809 c.201T>G p.P67P Chr.1: 100058686-100058686 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958571789 Chr.1: 100058781-100058781 on Build GRCh38
rs541959359 Chr.1: 100058707-100058707 on Build GRCh38
rs766111849 Chr.1: 100058689-100058692 on Build GRCh38
rs759483613 Chr.1: 100058767-100058772 on Build GRCh38
rs376137141 Chr.1: 100058624-100058624 on Build GRCh38
rs12119548 Chr.1: 100058780-100058780 on Build GRCh38
rs868036030 Chr.1: 100058589-100058589 on Build GRCh38
rs746873913 Chr.1: 100058757-100058757 on Build GRCh38
rs758400393 Chr.1: 100058711-100058711 on Build GRCh38
rs753466546 Chr.1: 100058636-100058636 on Build GRCh38
rs748718868 Chr.1: 100058758-100058758 on Build GRCh38
rs755690360 Chr.1: 100058634-100058634 on Build GRCh38
rs751480558 Chr.1: 100058761-100058764 on Build GRCh38
rs754446055 Chr.1: 100058638-100058638 on Build GRCh38
rs75771715 Chr.1: 100058670-100058670 on Build GRCh38
rs750883953 Chr.1: 100058629-100058629 on Build GRCh38
rs969184719 Chr.1: 100058614-100058614 on Build GRCh38
rs774928891 Chr.1: 100058616-100058616 on Build GRCh38
rs868164519 Chr.1: 100058709-100058709 on Build GRCh38
rs1026724812 Chr.1: 100058672-100058672 on Build GRCh38
rs770656529 Chr.1: 100058772-100058772 on Build GRCh38
rs749737937 Chr.1: 100058699-100058699 on Build GRCh38
rs777516977 Chr.1: 100058723-100058723 on Build GRCh38
rs1020434408 Chr.1: 100058592-100058592 on Build GRCh38
rs968536135 Chr.1: 100058613-100058613 on Build GRCh38
rs775738120 Chr.1: 100058775-100058775 on Build GRCh38
rs749473887 Chr.1: 100058777-100058777 on Build GRCh38
rs546293179 Chr.1: 100058666-100058666 on Build GRCh38
rs762419270 Chr.1: 100058620-100058620 on Build GRCh38
rs778409330 Chr.1: 100058651-100058651 on Build GRCh38

View Full Product Details