Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167546066-167546339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG1
Entrez Gene ID
8804
Gene Name
cellular repressor of E1A stimulated genes 1
Gene Aliases
CREG
UniGene
Hs.5710
Gene Chromosome Location
Chr.1: 167541013-167553819 on Build GRCh38
Transcripts
NM_003851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899086 c.622A>C p.I208L Chr.1: 167546138-167546138 on Build GRCh38
899087 c.520C>T p.R174* Chr.1: 167546240-167546240 on Build GRCh38
5599343 c.534G>A p.M178I Chr.1: 167546226-167546226 on Build GRCh38
6313957 c.653C>A p.T218K Chr.1: 167546107-167546107 on Build GRCh38
2185745 c.625G>A p.V209M Chr.1: 167546135-167546135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766343738 Chr.1: 167546173-167546173 on Build GRCh38
rs759385722 Chr.1: 167546186-167546186 on Build GRCh38
rs770079035 Chr.1: 167546161-167546161 on Build GRCh38
rs564738985 Chr.1: 167546112-167546114 on Build GRCh38
rs373686100 Chr.1: 167546191-167546191 on Build GRCh38
rs748480112 Chr.1: 167546214-167546214 on Build GRCh38
rs773453068 Chr.1: 167546169-167546169 on Build GRCh38
rs755385913 Chr.1: 167546210-167546210 on Build GRCh38
rs199570821 Chr.1: 167546224-167546224 on Build GRCh38
rs747612208 Chr.1: 167546208-167546208 on Build GRCh38
rs753556329 Chr.1: 167546286-167546286 on Build GRCh38
rs571629712 Chr.1: 167546250-167546250 on Build GRCh38
rs144088168 Chr.1: 167546113-167546113 on Build GRCh38
rs967967902 Chr.1: 167546115-167546115 on Build GRCh38
rs774281312 Chr.1: 167546240-167546240 on Build GRCh38
rs753468121 Chr.1: 167546199-167546199 on Build GRCh38
rs370827236 Chr.1: 167546135-167546135 on Build GRCh38
rs376218103 Chr.1: 167546136-167546136 on Build GRCh38
rs781696401 Chr.1: 167546213-167546213 on Build GRCh38
rs748398421 Chr.1: 167546152-167546152 on Build GRCh38
rs527573465 Chr.1: 167546094-167546094 on Build GRCh38
rs763688984 Chr.1: 167546198-167546198 on Build GRCh38
rs200055835 Chr.1: 167546188-167546188 on Build GRCh38
rs368150878 Chr.1: 167546206-167546206 on Build GRCh38
rs141237606 Chr.1: 167546118-167546118 on Build GRCh38
rs1031560799 Chr.1: 167546151-167546151 on Build GRCh38
rs775287487 Chr.1: 167546255-167546255 on Build GRCh38
rs763846546 Chr.1: 167546283-167546283 on Build GRCh38
rs371241698 Chr.1: 167546102-167546102 on Build GRCh38
rs756834421 Chr.1: 167546202-167546202 on Build GRCh38
rs757372798 Chr.1: 167546187-167546187 on Build GRCh38
rs376213841 Chr.1: 167546178-167546178 on Build GRCh38
rs764819478 Chr.1: 167546298-167546298 on Build GRCh38
rs183830710 Chr.1: 167546251-167546251 on Build GRCh38
rs755722100 Chr.1: 167546094-167546094 on Build GRCh38
rs763037302 Chr.1: 167546171-167546171 on Build GRCh38
rs777278355 Chr.1: 167546091-167546091 on Build GRCh38
rs749437307 Chr.1: 167546237-167546237 on Build GRCh38
rs771106885 Chr.1: 167546239-167546239 on Build GRCh38
rs763907354 Chr.1: 167546121-167546123 on Build GRCh38
rs748757226 Chr.1: 167546112-167546112 on Build GRCh38
rs368491115 Chr.1: 167546166-167546166 on Build GRCh38
rs770158928 Chr.1: 167546217-167546217 on Build GRCh38
rs760512764 Chr.1: 167546260-167546260 on Build GRCh38
rs761360132 Chr.1: 167546296-167546296 on Build GRCh38

View Full Product Details