Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43311844-43312083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5494427 c.245T>A p.M82K Chr.7: 43311980-43311980 on Build GRCh38
3366931 c.261C>A p.Y87* Chr.7: 43311996-43311996 on Build GRCh38
5662310 c.193C>T p.R65C Chr.7: 43311928-43311928 on Build GRCh38
1239051 c.113C>T p.P38L Chr.7: 43311848-43311848 on Build GRCh38
4542962 c.328G>A p.D110N Chr.7: 43312063-43312063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778885511 Chr.7: 43311882-43311882 on Build GRCh38
rs199717525 Chr.7: 43312032-43312032 on Build GRCh38
rs768048121 Chr.7: 43311901-43311901 on Build GRCh38
rs773660516 Chr.7: 43311940-43311940 on Build GRCh38
rs186064841 Chr.7: 43312057-43312057 on Build GRCh38
rs753946140 Chr.7: 43311874-43311874 on Build GRCh38
rs751138220 Chr.7: 43311962-43311963 on Build GRCh38
rs777589869 Chr.7: 43311905-43311905 on Build GRCh38
rs751383939 Chr.7: 43312020-43312020 on Build GRCh38
rs375404889 Chr.7: 43312056-43312056 on Build GRCh38
rs202230638 Chr.7: 43311949-43311949 on Build GRCh38
rs762360674 Chr.7: 43311931-43311931 on Build GRCh38
rs748099103 Chr.7: 43311885-43311885 on Build GRCh38
rs756332501 Chr.7: 43311862-43311862 on Build GRCh38
rs759666398 Chr.7: 43311975-43311975 on Build GRCh38
rs368244752 Chr.7: 43311959-43311959 on Build GRCh38
rs758391305 Chr.7: 43312002-43312002 on Build GRCh38
rs746740077 Chr.7: 43311910-43311910 on Build GRCh38
rs754051189 Chr.7: 43311974-43311974 on Build GRCh38
rs370846695 Chr.7: 43312015-43312015 on Build GRCh38
rs766605415 Chr.7: 43311963-43311963 on Build GRCh38
rs866226633 Chr.7: 43311871-43311871 on Build GRCh38
rs770727253 Chr.7: 43311916-43311916 on Build GRCh38
rs906767913 Chr.7: 43311909-43311909 on Build GRCh38
rs989967700 Chr.7: 43311980-43311980 on Build GRCh38
rs796167091 Chr.7: 43311896-43311896 on Build GRCh38
rs780933393 Chr.7: 43311925-43311925 on Build GRCh38
rs758173858 Chr.7: 43311891-43311891 on Build GRCh38
rs370542628 Chr.7: 43311900-43311900 on Build GRCh38
rs945355561 Chr.7: 43312039-43312039 on Build GRCh38
rs976128268 Chr.7: 43312011-43312011 on Build GRCh38
rs752614086 Chr.7: 43311993-43311993 on Build GRCh38
rs570717308 Chr.7: 43311978-43311978 on Build GRCh38
rs201089900 Chr.7: 43311935-43311935 on Build GRCh38
rs374211011 Chr.7: 43311928-43311928 on Build GRCh38
rs750970386 Chr.7: 43311929-43311929 on Build GRCh38
rs35544162 Chr.7: 43311896-43311896 on Build GRCh38
rs766550290 Chr.7: 43311866-43311866 on Build GRCh38
rs754996920 Chr.7: 43311881-43311881 on Build GRCh38

View Full Product Details