Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12084517-12084774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHTKD1
Entrez Gene ID
55526
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Gene Aliases
AMOXAD, CMT2Q
UniGene
Hs.104980
Gene Chromosome Location
Chr.10: 12068917-12123228 on Build GRCh38
Transcripts
NM_018706.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7191938 c.371A>C p.Q124P Chr.10: 12084600-12084600 on Build GRCh38
216915 c.435G>T p.W145C Chr.10: 12084664-12084664 on Build GRCh38
5431877 c.397G>C p.E133Q Chr.10: 12084626-12084626 on Build GRCh38
5012121 c.461A>G p.Q154R Chr.10: 12084690-12084690 on Build GRCh38
5094920 c.346G>T p.E116* Chr.10: 12084575-12084575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775948480 Chr.10: 12084700-12084700 on Build GRCh38
rs757610245 Chr.10: 12084676-12084676 on Build GRCh38
rs757387152 Chr.10: 12084726-12084726 on Build GRCh38
rs138219515 Chr.10: 12084730-12084730 on Build GRCh38
rs755902715 Chr.10: 12084632-12084632 on Build GRCh38
rs953186201 Chr.10: 12084649-12084649 on Build GRCh38
rs759872323 Chr.10: 12084555-12084555 on Build GRCh38
rs770329984 Chr.10: 12084636-12084639 on Build GRCh38
rs141676270 Chr.10: 12084672-12084672 on Build GRCh38
rs375917261 Chr.10: 12084699-12084699 on Build GRCh38
rs764537802 Chr.10: 12084568-12084568 on Build GRCh38
rs1030206721 Chr.10: 12084623-12084623 on Build GRCh38
rs908926249 Chr.10: 12084680-12084680 on Build GRCh38
rs746080416 Chr.10: 12084608-12084608 on Build GRCh38
rs369465951 Chr.10: 12084737-12084737 on Build GRCh38
rs767553524 Chr.10: 12084729-12084729 on Build GRCh38
rs754813064 Chr.10: 12084751-12084751 on Build GRCh38
rs35638771 Chr.10: 12084598-12084598 on Build GRCh38
rs377271668 Chr.10: 12084734-12084734 on Build GRCh38
rs765615698 Chr.10: 12084562-12084562 on Build GRCh38
rs897054680 Chr.10: 12084609-12084609 on Build GRCh38
rs763171314 Chr.10: 12084707-12084707 on Build GRCh38
rs200967740 Chr.10: 12084566-12084566 on Build GRCh38
rs773052090 Chr.10: 12084675-12084675 on Build GRCh38
rs779927813 Chr.10: 12084652-12084652 on Build GRCh38
rs781461520 Chr.10: 12084603-12084603 on Build GRCh38
rs555471579 Chr.10: 12084661-12084661 on Build GRCh38
rs772990698 Chr.10: 12084552-12084552 on Build GRCh38
rs369183622 Chr.10: 12084558-12084558 on Build GRCh38
rs78189904 Chr.10: 12084717-12084717 on Build GRCh38
rs141125831 Chr.10: 12084674-12084674 on Build GRCh38
rs185240347 Chr.10: 12084630-12084630 on Build GRCh38
rs764591027 Chr.10: 12084716-12084716 on Build GRCh38
rs757261214 Chr.10: 12084599-12084599 on Build GRCh38

View Full Product Details