Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49972948-49973197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX4
Entrez Gene ID
1748
Gene Name
distal-less homeobox 4
Gene Aliases
BP1, DLX7, DLX8, DLX9, OFC15
UniGene
Hs.591167
Gene Chromosome Location
Chr.17: 49968970-49974959 on Build GRCh38
Transcripts
NM_001934.3 NM_138281.2 XM_017024291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6645122 c.106C>T p.R36W Chr.17: 49973111-49973111 on Build GRCh38
3889859 c.102C>T p.S34S Chr.17: 49973107-49973107 on Build GRCh38
6645121 c.322C>T p.R108W Chr.17: 49973111-49973111 on Build GRCh38
3889858 c.318C>T p.S106S Chr.17: 49973107-49973107 on Build GRCh38
706997 c.319G>T p.E107* Chr.17: 49973108-49973108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749670037 Chr.17: 49973114-49973114 on Build GRCh38
rs751534594 Chr.17: 49973027-49973027 on Build GRCh38
rs370138317 Chr.17: 49973131-49973132 on Build GRCh38
rs765117074 Chr.17: 49973006-49973006 on Build GRCh38
rs755026052 Chr.17: 49973102-49973102 on Build GRCh38
rs752997759 Chr.17: 49973108-49973108 on Build GRCh38
rs763724785 Chr.17: 49972978-49972978 on Build GRCh38
rs774177303 Chr.17: 49973146-49973146 on Build GRCh38
rs748457781 Chr.17: 49973034-49973034 on Build GRCh38
rs754160098 Chr.17: 49973175-49973175 on Build GRCh38
rs767672602 Chr.17: 49973158-49973158 on Build GRCh38
rs769494752 Chr.17: 49973139-49973139 on Build GRCh38
rs766449181 Chr.17: 49973017-49973017 on Build GRCh38
rs779305699 Chr.17: 49973120-49973120 on Build GRCh38
rs776348139 Chr.17: 49972997-49972997 on Build GRCh38
rs756579600 Chr.17: 49973036-49973036 on Build GRCh38
rs746562515 Chr.17: 49973049-49973049 on Build GRCh38
rs751656882 Chr.17: 49973021-49973021 on Build GRCh38
rs778257713 Chr.17: 49973113-49973113 on Build GRCh38
rs902773730 Chr.17: 49973132-49973132 on Build GRCh38
rs187087248 Chr.17: 49972970-49972970 on Build GRCh38
rs755214946 Chr.17: 49973025-49973025 on Build GRCh38
rs768188135 Chr.17: 49973124-49973124 on Build GRCh38
rs764473165 Chr.17: 49973173-49973173 on Build GRCh38
rs768284734 Chr.17: 49973053-49973053 on Build GRCh38
rs759620239 Chr.17: 49973150-49973150 on Build GRCh38
rs760963288 Chr.17: 49973160-49973160 on Build GRCh38
rs370757334 Chr.17: 49973024-49973024 on Build GRCh38
rs771385999 Chr.17: 49973046-49973046 on Build GRCh38
rs750380500 Chr.17: 49973014-49973014 on Build GRCh38
rs762935432 Chr.17: 49973089-49973089 on Build GRCh38
rs766294623 Chr.17: 49973096-49973096 on Build GRCh38
rs757773828 Chr.17: 49973117-49973117 on Build GRCh38
rs575249063 Chr.17: 49973111-49973111 on Build GRCh38
rs749783136 Chr.17: 49973042-49973042 on Build GRCh38
rs1013743575 Chr.17: 49973070-49973070 on Build GRCh38
rs778378113 Chr.17: 49973037-49973037 on Build GRCh38
rs761559100 Chr.17: 49973073-49973073 on Build GRCh38
rs940042366 Chr.17: 49972974-49972974 on Build GRCh38
rs758314831 Chr.17: 49973015-49973015 on Build GRCh38
rs555311434 Chr.17: 49973061-49973061 on Build GRCh38
rs943084049 Chr.17: 49973071-49973071 on Build GRCh38
rs868538642 Chr.17: 49973087-49973087 on Build GRCh38
rs780722446 Chr.17: 49973129-49973129 on Build GRCh38
rs773041429 Chr.17: 49973086-49973086 on Build GRCh38
rs775064973 Chr.17: 49973047-49973047 on Build GRCh38
rs780703647 Chr.17: 49973016-49973026 on Build GRCh38
rs764988416 Chr.17: 49973076-49973076 on Build GRCh38
rs1018429591 Chr.17: 49973101-49973101 on Build GRCh38
rs767797609 Chr.17: 49973104-49973104 on Build GRCh38
rs746483420 Chr.17: 49973123-49973123 on Build GRCh38
rs762661270 Chr.17: 49973143-49973143 on Build GRCh38
rs770843158 Chr.17: 49973144-49973144 on Build GRCh38
rs989434221 Chr.17: 49972994-49972994 on Build GRCh38
rs372156240 Chr.17: 49973142-49973142 on Build GRCh38
rs749719388 Chr.17: 49973129-49973129 on Build GRCh38
rs747779313 Chr.17: 49973131-49973131 on Build GRCh38
rs1048145603 Chr.17: 49973157-49973157 on Build GRCh38
rs901469869 Chr.17: 49973097-49973097 on Build GRCh38

View Full Product Details