Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42835593-42835850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMAP
Entrez Gene ID
114625
Gene Name
erythroblast membrane associated protein (Scianna blood group)
Gene Aliases
BTN5, PRO2801, RD, SC
UniGene
Hs.439437
Gene Chromosome Location
Chr.1: 42817105-42844991 on Build GRCh38
Transcripts
NM_001017922.1 NM_018538.3 XM_006710313.3 XM_011540570.2 XM_011540571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681652 c.575C>T p.T192M Chr.1: 42835756-42835756 on Build GRCh38
6656183 c.576G>A p.T192T Chr.1: 42835757-42835757 on Build GRCh38
6227554 c.563A>G p.Y188C Chr.1: 42835744-42835744 on Build GRCh38
5637912 c.576G>T p.T192T Chr.1: 42835757-42835757 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753063024 Chr.1: 42835782-42835783 on Build GRCh38
rs768305324 Chr.1: 42835750-42835750 on Build GRCh38
rs774598319 Chr.1: 42835714-42835715 on Build GRCh38
rs745766742 Chr.1: 42835791-42835791 on Build GRCh38
rs757267796 Chr.1: 42835693-42835693 on Build GRCh38
rs866116043 Chr.1: 42835762-42835762 on Build GRCh38
rs956366886 Chr.1: 42835656-42835656 on Build GRCh38
rs35767222 Chr.1: 42835822-42835822 on Build GRCh38
rs574092922 Chr.1: 42835621-42835621 on Build GRCh38
rs1024642297 Chr.1: 42835691-42835691 on Build GRCh38
rs771683714 Chr.1: 42835734-42835734 on Build GRCh38
rs149218926 Chr.1: 42835827-42835828 on Build GRCh38
rs775244639 Chr.1: 42835736-42835736 on Build GRCh38
rs201485804 Chr.1: 42835784-42835784 on Build GRCh38
rs764314084 Chr.1: 42835782-42835782 on Build GRCh38
rs746668638 Chr.1: 42835742-42835742 on Build GRCh38
rs1009226888 Chr.1: 42835675-42835675 on Build GRCh38
rs113445278 Chr.1: 42835756-42835756 on Build GRCh38
rs367947427 Chr.1: 42835764-42835764 on Build GRCh38
rs761130083 Chr.1: 42835788-42835788 on Build GRCh38
rs34475455 Chr.1: 42835783-42835783 on Build GRCh38
rs374232709 Chr.1: 42835780-42835780 on Build GRCh38
rs371707071 Chr.1: 42835765-42835765 on Build GRCh38
rs748010417 Chr.1: 42835771-42835772 on Build GRCh38
rs749038102 Chr.1: 42835681-42835681 on Build GRCh38
rs779040282 Chr.1: 42835785-42835785 on Build GRCh38
rs756306834 Chr.1: 42835769-42835769 on Build GRCh38
rs139202900 Chr.1: 42835618-42835618 on Build GRCh38
rs759891051 Chr.1: 42835726-42835726 on Build GRCh38
rs767928266 Chr.1: 42835781-42835781 on Build GRCh38
rs758311333 Chr.1: 42835806-42835806 on Build GRCh38
rs943517600 Chr.1: 42835821-42835822 on Build GRCh38
rs1032369944 Chr.1: 42835628-42835628 on Build GRCh38
rs374399714 Chr.1: 42835757-42835757 on Build GRCh38
rs149950197 Chr.1: 42835720-42835720 on Build GRCh38
rs370086635 Chr.1: 42835739-42835739 on Build GRCh38
rs144054863 Chr.1: 42835760-42835760 on Build GRCh38
rs922651477 Chr.1: 42835825-42835825 on Build GRCh38
rs778993311 Chr.1: 42835700-42835700 on Build GRCh38
rs542726162 Chr.1: 42835670-42835670 on Build GRCh38
rs779449270 Chr.1: 42835812-42835812 on Build GRCh38

View Full Product Details