Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127582130-127582391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
UniGene
Hs.521168
Gene Chromosome Location
Chr.7: 127580628-127585600 on Build GRCh38
Transcripts
NM_024523.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3942009 c.2130C>A p.I710I Chr.7: 127582212-127582212 on Build GRCh38
5434747 c.2179A>G p.I727V Chr.7: 127582163-127582163 on Build GRCh38
325392 c.1997A>T p.E666V Chr.7: 127582345-127582345 on Build GRCh38
1085129 c.2199A>G p.L733L Chr.7: 127582143-127582143 on Build GRCh38
331530 c.2032G>T p.E678* Chr.7: 127582310-127582310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778788276 Chr.7: 127582203-127582203 on Build GRCh38
rs986532186 Chr.7: 127582252-127582252 on Build GRCh38
rs766103716 Chr.7: 127582265-127582265 on Build GRCh38
rs146301759 Chr.7: 127582256-127582256 on Build GRCh38
rs1002952037 Chr.7: 127582360-127582360 on Build GRCh38
rs759635348 Chr.7: 127582270-127582270 on Build GRCh38
rs758548110 Chr.7: 127582285-127582285 on Build GRCh38
rs886922070 Chr.7: 127582247-127582247 on Build GRCh38
rs765477912 Chr.7: 127582272-127582272 on Build GRCh38
rs755237786 Chr.7: 127582200-127582200 on Build GRCh38
rs778594207 Chr.7: 127582313-127582313 on Build GRCh38
rs141951933 Chr.7: 127582224-127582224 on Build GRCh38
rs747860732 Chr.7: 127582333-127582333 on Build GRCh38
rs753989772 Chr.7: 127582198-127582198 on Build GRCh38
rs767463630 Chr.7: 127582174-127582174 on Build GRCh38
rs370136814 Chr.7: 127582246-127582246 on Build GRCh38
rs200271393 Chr.7: 127582358-127582358 on Build GRCh38
rs750408956 Chr.7: 127582189-127582189 on Build GRCh38
rs781374297 Chr.7: 127582305-127582305 on Build GRCh38
rs771608741 Chr.7: 127582260-127582260 on Build GRCh38
rs370968860 Chr.7: 127582326-127582326 on Build GRCh38
rs773918151 Chr.7: 127582157-127582157 on Build GRCh38
rs200409039 Chr.7: 127582173-127582173 on Build GRCh38
rs771413693 Chr.7: 127582226-127582226 on Build GRCh38
rs746269017 Chr.7: 127582308-127582308 on Build GRCh38
rs148336273 Chr.7: 127582223-127582223 on Build GRCh38
rs768387408 Chr.7: 127582241-127582241 on Build GRCh38
rs753671181 Chr.7: 127582266-127582266 on Build GRCh38
rs770102564 Chr.7: 127582312-127582312 on Build GRCh38
rs757848124 Chr.7: 127582304-127582304 on Build GRCh38
rs911089387 Chr.7: 127582254-127582254 on Build GRCh38
rs374924542 Chr.7: 127582191-127582191 on Build GRCh38
rs147718839 Chr.7: 127582341-127582341 on Build GRCh38
rs752904334 Chr.7: 127582283-127582283 on Build GRCh38
rs773682905 Chr.7: 127582255-127582255 on Build GRCh38
rs765009526 Chr.7: 127582359-127582359 on Build GRCh38
rs748826945 Chr.7: 127582227-127582227 on Build GRCh38
rs777726921 Chr.7: 127582231-127582231 on Build GRCh38
rs375417520 Chr.7: 127582347-127582347 on Build GRCh38
rs746355709 Chr.7: 127582256-127582257 on Build GRCh38
rs73234845 Chr.7: 127582264-127582264 on Build GRCh38
rs375182497 Chr.7: 127582156-127582156 on Build GRCh38
rs776237823 Chr.7: 127582357-127582357 on Build GRCh38
rs770930978 Chr.7: 127582354-127582354 on Build GRCh38
rs778057410 Chr.7: 127582298-127582298 on Build GRCh38
rs374318153 Chr.7: 127582262-127582262 on Build GRCh38
rs780756357 Chr.7: 127582189-127582199 on Build GRCh38
rs752059605 Chr.7: 127582299-127582299 on Build GRCh38
rs760539071 Chr.7: 127582351-127582351 on Build GRCh38
rs371762826 Chr.7: 127582164-127582164 on Build GRCh38
rs752851618 Chr.7: 127582204-127582204 on Build GRCh38
rs369087622 Chr.7: 127582215-127582215 on Build GRCh38
rs752951831 Chr.7: 127582365-127582365 on Build GRCh38

View Full Product Details