Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21502999-21503230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU2
Entrez Gene ID
221154
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
1110008L20Rik, EFHA1
UniGene
Hs.412103
Gene Chromosome Location
Chr.13: 21492689-21604216 on Build GRCh38
Transcripts
NM_152726.2 XM_017020433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945972 c.811T>C p.F271L Chr.13: 21503048-21503048 on Build GRCh38
4680316 c.832A>G p.M278V Chr.13: 21503027-21503027 on Build GRCh38
6905230 c.842A>G p.E281G Chr.13: 21503017-21503017 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767707249 Chr.13: 21503039-21503040 on Build GRCh38
rs752960720 Chr.13: 21503072-21503073 on Build GRCh38
rs17853349 Chr.13: 21503080-21503080 on Build GRCh38
rs751370417 Chr.13: 21503061-21503061 on Build GRCh38
rs956656900 Chr.13: 21503103-21503103 on Build GRCh38
rs766517159 Chr.13: 21503037-21503037 on Build GRCh38
rs765534360 Chr.13: 21503049-21503049 on Build GRCh38
rs368352095 Chr.13: 21503025-21503025 on Build GRCh38
rs376598541 Chr.13: 21503162-21503162 on Build GRCh38
rs111829892 Chr.13: 21503194-21503194 on Build GRCh38
rs967833958 Chr.13: 21503034-21503034 on Build GRCh38
rs754144412 Chr.13: 21503043-21503043 on Build GRCh38
rs182607636 Chr.13: 21503134-21503134 on Build GRCh38
rs757072057 Chr.13: 21503086-21503086 on Build GRCh38
rs374979910 Chr.13: 21503128-21503128 on Build GRCh38
rs760848974 Chr.13: 21503031-21503031 on Build GRCh38
rs773613035 Chr.13: 21503124-21503124 on Build GRCh38
rs748771205 Chr.13: 21503107-21503107 on Build GRCh38
rs575134900 Chr.13: 21503168-21503168 on Build GRCh38
rs759675299 Chr.13: 21503129-21503129 on Build GRCh38
rs992233507 Chr.13: 21503115-21503115 on Build GRCh38
rs779502900 Chr.13: 21503102-21503102 on Build GRCh38
rs774045988 Chr.13: 21503113-21503113 on Build GRCh38
rs748463277 Chr.13: 21503114-21503114 on Build GRCh38
rs755648270 Chr.13: 21503092-21503092 on Build GRCh38
rs1039256973 Chr.13: 21503172-21503172 on Build GRCh38
rs761223512 Chr.13: 21503126-21503126 on Build GRCh38
rs565422565 Chr.13: 21503087-21503087 on Build GRCh38
rs745763930 Chr.13: 21503090-21503090 on Build GRCh38
rs900359445 Chr.13: 21503188-21503188 on Build GRCh38
rs765445844 Chr.13: 21503133-21503133 on Build GRCh38
rs566143584 Chr.13: 21503159-21503159 on Build GRCh38
rs781745977 Chr.13: 21503130-21503130 on Build GRCh38
rs532741389 Chr.13: 21503111-21503111 on Build GRCh38
rs772422859 Chr.13: 21503122-21503122 on Build GRCh38

View Full Product Details