Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44506374-44506623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCB
Entrez Gene ID
84075
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
C14orf155
UniGene
Hs.307086
Gene Chromosome Location
Chr.14: 44504149-44507296 on Build GRCh38
Transcripts
NM_032135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4822075 c.485C>G p.S162* Chr.14: 44506503-44506503 on Build GRCh38
6667431 c.417C>T p.Y139Y Chr.14: 44506571-44506571 on Build GRCh38
6305150 c.560C>A p.S187* Chr.14: 44506428-44506428 on Build GRCh38
4499486 c.543C>T p.A181A Chr.14: 44506445-44506445 on Build GRCh38
337004 c.459_460GG>TC p.K153>? Chr.14: 44506528-44506529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560077187 Chr.14: 44506436-44506436 on Build GRCh38
rs181538481 Chr.14: 44506534-44506534 on Build GRCh38
rs1000733264 Chr.14: 44506540-44506540 on Build GRCh38
rs201893311 Chr.14: 44506590-44506590 on Build GRCh38
rs150383523 Chr.14: 44506484-44506484 on Build GRCh38
rs201253705 Chr.14: 44506525-44506525 on Build GRCh38
rs369074703 Chr.14: 44506582-44506582 on Build GRCh38
rs527611726 Chr.14: 44506447-44506447 on Build GRCh38
rs199704834 Chr.14: 44506408-44506408 on Build GRCh38
rs760879258 Chr.14: 44506443-44506443 on Build GRCh38
rs752491125 Chr.14: 44506588-44506588 on Build GRCh38
rs140542441 Chr.14: 44506513-44506513 on Build GRCh38
rs3809429 Chr.14: 44506403-44506403 on Build GRCh38
rs879203791 Chr.14: 44506439-44506439 on Build GRCh38
rs867061377 Chr.14: 44506497-44506497 on Build GRCh38
rs772280580 Chr.14: 44506510-44506510 on Build GRCh38
rs199524512 Chr.14: 44506532-44506532 on Build GRCh38
rs779299842 Chr.14: 44506492-44506492 on Build GRCh38
rs114872568 Chr.14: 44506568-44506568 on Build GRCh38
rs200905673 Chr.14: 44506414-44506414 on Build GRCh38
rs745883410 Chr.14: 44506501-44506501 on Build GRCh38
rs564980791 Chr.14: 44506458-44506458 on Build GRCh38
rs772868501 Chr.14: 44506554-44506554 on Build GRCh38
rs371906913 Chr.14: 44506459-44506459 on Build GRCh38
rs761835952 Chr.14: 44506472-44506474 on Build GRCh38
rs760903043 Chr.14: 44506401-44506401 on Build GRCh38
rs757929406 Chr.14: 44506461-44506461 on Build GRCh38
rs758303849 Chr.14: 44506397-44506397 on Build GRCh38
rs755985760 Chr.14: 44506589-44506589 on Build GRCh38
rs776077068 Chr.14: 44506442-44506442 on Build GRCh38
rs532380844 Chr.14: 44506483-44506483 on Build GRCh38
rs780913296 Chr.14: 44506419-44506419 on Build GRCh38
rs201940751 Chr.14: 44506515-44506515 on Build GRCh38
rs531498905 Chr.14: 44506467-44506467 on Build GRCh38
rs188430266 Chr.14: 44506581-44506581 on Build GRCh38
rs552049784 Chr.14: 44506455-44506455 on Build GRCh38
rs1018854001 Chr.14: 44506465-44506465 on Build GRCh38
rs764824148 Chr.14: 44506551-44506551 on Build GRCh38
rs746590611 Chr.14: 44506466-44506468 on Build GRCh38
rs780002065 Chr.14: 44506402-44506402 on Build GRCh38
rs766313987 Chr.14: 44506487-44506489 on Build GRCh38
rs754213779 Chr.14: 44506448-44506448 on Build GRCh38
rs542027117 Chr.14: 44506427-44506427 on Build GRCh38
rs34582231 Chr.14: 44506557-44506557 on Build GRCh38
rs754925824 Chr.14: 44506479-44506479 on Build GRCh38
rs762385492 Chr.14: 44506555-44506555 on Build GRCh38
rs771237972 Chr.14: 44506488-44506488 on Build GRCh38
rs746660396 Chr.14: 44506428-44506428 on Build GRCh38
rs751564957 Chr.14: 44506554-44506555 on Build GRCh38
rs531702157 Chr.14: 44506449-44506449 on Build GRCh38
rs779801157 Chr.14: 44506462-44506462 on Build GRCh38
rs768816708 Chr.14: 44506414-44506415 on Build GRCh38

View Full Product Details