Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32937317-32937542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMB
Entrez Gene ID
3109
Gene Name
major histocompatibility complex, class II, DM beta
Gene Aliases
D6S221E, RING7
UniGene
Hs.351279
Gene Chromosome Location
Chr.6: 32934629-32941070 on Build GRCh38
Transcripts
NM_002118.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624004 c.366C>T p.T122T Chr.6: 32937428-32937428 on Build GRCh38
6680770 c.406T>C p.Y136H Chr.6: 32937388-32937388 on Build GRCh38
742395 c.366C>A p.T122T Chr.6: 32937428-32937428 on Build GRCh38
742397 c.381G>T p.T127T Chr.6: 32937413-32937413 on Build GRCh38
5624005 c.366C>T p.T122T Chr.6: 32937428-32937428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145745508 Chr.6: 32937350-32937350 on Build GRCh38
rs752219540 Chr.6: 32937449-32937449 on Build GRCh38
rs759898785 Chr.6: 32937437-32937437 on Build GRCh38
rs745762577 Chr.6: 32937380-32937380 on Build GRCh38
rs372328837 Chr.6: 32937413-32937413 on Build GRCh38
rs745576215 Chr.6: 32937359-32937359 on Build GRCh38
rs772240569 Chr.6: 32937406-32937406 on Build GRCh38
rs746861381 Chr.6: 32937351-32937351 on Build GRCh38
rs148929307 Chr.6: 32937353-32937353 on Build GRCh38
rs768804217 Chr.6: 32937392-32937392 on Build GRCh38
rs995890905 Chr.6: 32937428-32937428 on Build GRCh38
rs916346407 Chr.6: 32937492-32937492 on Build GRCh38
rs777141527 Chr.6: 32937435-32937435 on Build GRCh38
rs1044113265 Chr.6: 32937376-32937376 on Build GRCh38
rs763706187 Chr.6: 32937506-32937506 on Build GRCh38
rs761285537 Chr.6: 32937467-32937468 on Build GRCh38
rs761066493 Chr.6: 32937425-32937425 on Build GRCh38
rs143633244 Chr.6: 32937395-32937395 on Build GRCh38
rs757914913 Chr.6: 32937471-32937471 on Build GRCh38
rs766687161 Chr.6: 32937429-32937429 on Build GRCh38
rs781321450 Chr.6: 32937375-32937375 on Build GRCh38
rs764811408 Chr.6: 32937442-32937442 on Build GRCh38
rs375089997 Chr.6: 32937511-32937511 on Build GRCh38
rs774238503 Chr.6: 32937393-32937393 on Build GRCh38
rs867799361 Chr.6: 32937414-32937414 on Build GRCh38
rs111943650 Chr.6: 32937426-32937426 on Build GRCh38
rs368649736 Chr.6: 32937342-32937342 on Build GRCh38

View Full Product Details