Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 96884673-96884942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH2
Entrez Gene ID
1730
Gene Name
diaphanous related formin 2
Gene Aliases
DIA, DIA2, DRF2, POF, POF2
UniGene
Hs.226483
Gene Chromosome Location
Chr.X: 96684663-97600598 on Build GRCh38
Transcripts
NM_006729.4 NM_007309.3 NM_013347.4

Gene Symbol
RPA4
Entrez Gene ID
29935
Gene Name
replication protein A4
Gene Aliases
HSU24186
UniGene
Hs.659349
Gene Chromosome Location
Chr.X: 96883908-96885467 on Build GRCh38
Transcripts
NM_006729.4 NM_007309.3 NM_013347.4

SNP Detail

SNP ID Chromosome Location
rs773964117 Chr.X: 96884765-96884765 on Build GRCh38
rs773158242 Chr.X: 96884780-96884780 on Build GRCh38
rs61736055 Chr.X: 96884879-96884879 on Build GRCh38
rs769490765 Chr.X: 96884751-96884751 on Build GRCh38
rs761871309 Chr.X: 96884775-96884775 on Build GRCh38
rs752735862 Chr.X: 96884825-96884825 on Build GRCh38
rs192760986 Chr.X: 96884782-96884782 on Build GRCh38
rs377302642 Chr.X: 96884705-96884705 on Build GRCh38
rs938173282 Chr.X: 96884824-96884824 on Build GRCh38
rs772143522 Chr.X: 96884896-96884896 on Build GRCh38
rs865958206 Chr.X: 96884734-96884734 on Build GRCh38
rs200380831 Chr.X: 96884878-96884878 on Build GRCh38
rs754066368 Chr.X: 96884793-96884793 on Build GRCh38
rs184130356 Chr.X: 96884798-96884798 on Build GRCh38
rs867176116 Chr.X: 96884872-96884872 on Build GRCh38
rs558028763 Chr.X: 96884915-96884915 on Build GRCh38
rs775071657 Chr.X: 96884752-96884752 on Build GRCh38
rs150547120 Chr.X: 96884747-96884747 on Build GRCh38
rs748929447 Chr.X: 96884881-96884881 on Build GRCh38
rs1029496439 Chr.X: 96884702-96884702 on Build GRCh38
rs187756303 Chr.X: 96884726-96884726 on Build GRCh38
rs768311636 Chr.X: 96884888-96884888 on Build GRCh38
rs139569203 Chr.X: 96884776-96884776 on Build GRCh38
rs373048681 Chr.X: 96884902-96884902 on Build GRCh38
rs367690619 Chr.X: 96884772-96884772 on Build GRCh38
rs778143589 Chr.X: 96884735-96884735 on Build GRCh38
rs906667923 Chr.X: 96884811-96884811 on Build GRCh38
rs201616178 Chr.X: 96884764-96884764 on Build GRCh38
rs778586646 Chr.X: 96884889-96884889 on Build GRCh38
rs759709110 Chr.X: 96884918-96884918 on Build GRCh38
rs765342276 Chr.X: 96884802-96884802 on Build GRCh38
rs1005836154 Chr.X: 96884900-96884900 on Build GRCh38
rs766197285 Chr.X: 96884792-96884792 on Build GRCh38
rs758918264 Chr.X: 96884851-96884851 on Build GRCh38
rs777899567 Chr.X: 96884850-96884851 on Build GRCh38
rs778184711 Chr.X: 96884873-96884873 on Build GRCh38
rs148408114 Chr.X: 96884717-96884717 on Build GRCh38
rs755083419 Chr.X: 96884710-96884710 on Build GRCh38
rs183526208 Chr.X: 96884720-96884720 on Build GRCh38
rs747607202 Chr.X: 96884894-96884894 on Build GRCh38
rs199631132 Chr.X: 96884903-96884903 on Build GRCh38
rs748956801 Chr.X: 96884754-96884754 on Build GRCh38
rs61752459 Chr.X: 96884730-96884730 on Build GRCh38
rs954018134 Chr.X: 96884721-96884721 on Build GRCh38

View Full Product Details