Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47811870-47812111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf69
Entrez Gene ID
80099
Gene Name
chromosome 7 open reading frame 69
Gene Aliases
-
UniGene
Hs.287647
Gene Chromosome Location
Chr.7: 47795291-47819847 on Build GRCh38
Transcripts
NM_001302627.1 NM_025031.2 NM_138295.3 XM_017011798.1

Gene Symbol
PKD1L1
Entrez Gene ID
168507
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
Gene Aliases
PRO19563
UniGene
Hs.195979
Gene Chromosome Location
Chr.7: 47774603-47958622 on Build GRCh38
Transcripts
NM_001302627.1 NM_025031.2 NM_138295.3 XM_017011798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6571090 c.7511C>T p.S2504L Chr.7: 47811887-47811887 on Build GRCh38
5813131 c.239T>C p.V80A Chr.7: 47811930-47811930 on Build GRCh38
5813130 c.7468T>C p.S2490P Chr.7: 47811930-47811930 on Build GRCh38
3881395 c.7493C>T p.T2498M Chr.7: 47811905-47811905 on Build GRCh38
79015 c.7511C>A p.S2504* Chr.7: 47811887-47811887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765051748 Chr.7: 47812071-47812071 on Build GRCh38
rs556770278 Chr.7: 47812027-47812027 on Build GRCh38
rs1000336857 Chr.7: 47812044-47812044 on Build GRCh38
rs73692834 Chr.7: 47812025-47812025 on Build GRCh38
rs781337672 Chr.7: 47811983-47811983 on Build GRCh38
rs761633022 Chr.7: 47811944-47811944 on Build GRCh38
rs767901351 Chr.7: 47811999-47811999 on Build GRCh38
rs774795639 Chr.7: 47811916-47811916 on Build GRCh38
rs1006144078 Chr.7: 47812005-47812005 on Build GRCh38
rs574494237 Chr.7: 47811988-47811988 on Build GRCh38
rs369536558 Chr.7: 47812051-47812051 on Build GRCh38
rs376886746 Chr.7: 47811998-47811998 on Build GRCh38
rs201532761 Chr.7: 47811933-47811933 on Build GRCh38
rs749817008 Chr.7: 47812004-47812004 on Build GRCh38
rs769091129 Chr.7: 47811906-47811906 on Build GRCh38
rs771280105 Chr.7: 47812011-47812011 on Build GRCh38
rs747289738 Chr.7: 47811892-47811892 on Build GRCh38
rs367611487 Chr.7: 47811905-47811905 on Build GRCh38
rs760384390 Chr.7: 47811919-47811919 on Build GRCh38
rs960164385 Chr.7: 47812078-47812078 on Build GRCh38
rs766485639 Chr.7: 47811962-47811962 on Build GRCh38
rs751536167 Chr.7: 47811974-47811974 on Build GRCh38
rs750386642 Chr.7: 47811946-47811946 on Build GRCh38
rs371672840 Chr.7: 47811934-47811934 on Build GRCh38
rs1028674672 Chr.7: 47811981-47811981 on Build GRCh38
rs373951831 Chr.7: 47812081-47812081 on Build GRCh38
rs1049158429 Chr.7: 47811890-47811890 on Build GRCh38
rs770279635 Chr.7: 47811895-47811895 on Build GRCh38
rs748357359 Chr.7: 47811984-47811984 on Build GRCh38
rs764996758 Chr.7: 47811945-47811945 on Build GRCh38
rs553682583 Chr.7: 47811904-47811904 on Build GRCh38
rs73103467 Chr.7: 47812056-47812056 on Build GRCh38
rs140456142 Chr.7: 47811980-47811980 on Build GRCh38
rs937199556 Chr.7: 47811931-47811931 on Build GRCh38
rs534330206 Chr.7: 47811958-47811958 on Build GRCh38
rs370071576 Chr.7: 47812073-47812073 on Build GRCh38
rs536060017 Chr.7: 47812037-47812037 on Build GRCh38
rs901619999 Chr.7: 47811969-47811969 on Build GRCh38
rs766461568 Chr.7: 47812065-47812065 on Build GRCh38
rs373762740 Chr.7: 47811897-47811897 on Build GRCh38
rs371450996 Chr.7: 47811894-47811894 on Build GRCh38
rs762891803 Chr.7: 47812085-47812085 on Build GRCh38
rs377233899 Chr.7: 47812063-47812063 on Build GRCh38

View Full Product Details