Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41699988-41700235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBA
Entrez Gene ID
3624
Gene Name
inhibin beta A subunit
Gene Aliases
EDF, FRP
UniGene
Hs.28792
Gene Chromosome Location
Chr.7: 41685101-41710532 on Build GRCh38
Transcripts
NM_002192.2 XM_017012174.1 XM_017012175.1 XM_017012176.1 NR_027118.1 NR_027119.1

Gene Symbol
INHBA-AS1
Entrez Gene ID
285954
Gene Name
INHBA antisense RNA 1
Gene Aliases
-
UniGene
Hs.656869
Gene Chromosome Location
Chr.7: 41693916-41779378 on Build GRCh38
Transcripts
NM_002192.2 XM_017012174.1 XM_017012175.1 XM_017012176.1 NR_027118.1 NR_027119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832779 c.164T>G p.V55G Chr.7: 41700211-41700211 on Build GRCh38
6934229 c.382G>A p.E128K Chr.7: 41699993-41699993 on Build GRCh38
1450705 c.347T>G p.L116R Chr.7: 41700028-41700028 on Build GRCh38
6177881 c.242C>A p.A81E Chr.7: 41700133-41700133 on Build GRCh38
6936112 c.147C>G p.N49K Chr.7: 41700228-41700228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756786763 Chr.7: 41700189-41700189 on Build GRCh38
rs756693243 Chr.7: 41700114-41700114 on Build GRCh38
rs780556409 Chr.7: 41700117-41700117 on Build GRCh38
rs749750926 Chr.7: 41700118-41700118 on Build GRCh38
rs760840635 Chr.7: 41700140-41700140 on Build GRCh38
rs368955824 Chr.7: 41700090-41700090 on Build GRCh38
rs771076354 Chr.7: 41700143-41700143 on Build GRCh38
rs763814285 Chr.7: 41700159-41700159 on Build GRCh38
rs748436551 Chr.7: 41700128-41700128 on Build GRCh38
rs778711643 Chr.7: 41700016-41700016 on Build GRCh38
rs770345466 Chr.7: 41700027-41700027 on Build GRCh38
rs746480596 Chr.7: 41700026-41700026 on Build GRCh38
rs751150931 Chr.7: 41700105-41700105 on Build GRCh38
rs780644703 Chr.7: 41700192-41700192 on Build GRCh38
rs755524019 Chr.7: 41700207-41700207 on Build GRCh38
rs779371975 Chr.7: 41700121-41700121 on Build GRCh38
rs768954945 Chr.7: 41700038-41700038 on Build GRCh38
rs975745915 Chr.7: 41700179-41700179 on Build GRCh38
rs753760792 Chr.7: 41700084-41700084 on Build GRCh38
rs752403730 Chr.7: 41700092-41700092 on Build GRCh38
rs776731486 Chr.7: 41700149-41700149 on Build GRCh38
rs773300358 Chr.7: 41700138-41700138 on Build GRCh38
rs569429371 Chr.7: 41700057-41700057 on Build GRCh38
rs749899848 Chr.7: 41700194-41700194 on Build GRCh38
rs757877629 Chr.7: 41700021-41700021 on Build GRCh38
rs551268437 Chr.7: 41700053-41700053 on Build GRCh38
rs780538034 Chr.7: 41700029-41700029 on Build GRCh38
rs139451450 Chr.7: 41700030-41700030 on Build GRCh38
rs1030538647 Chr.7: 41700213-41700213 on Build GRCh38
rs777279223 Chr.7: 41700103-41700103 on Build GRCh38
rs540086083 Chr.7: 41700086-41700086 on Build GRCh38
rs774463919 Chr.7: 41700042-41700042 on Build GRCh38
rs773386945 Chr.7: 41700077-41700077 on Build GRCh38
rs1001765662 Chr.7: 41700055-41700055 on Build GRCh38
rs182017908 Chr.7: 41700151-41700151 on Build GRCh38
rs751118764 Chr.7: 41700160-41700160 on Build GRCh38
rs1019880841 Chr.7: 41700144-41700144 on Build GRCh38
rs747778899 Chr.7: 41700020-41700020 on Build GRCh38
rs919035995 Chr.7: 41700201-41700201 on Build GRCh38
rs777383877 Chr.7: 41700025-41700025 on Build GRCh38
rs772394193 Chr.7: 41700129-41700129 on Build GRCh38
rs752596382 Chr.7: 41700152-41700152 on Build GRCh38
rs769036465 Chr.7: 41700120-41700120 on Build GRCh38
rs757935652 Chr.7: 41700075-41700075 on Build GRCh38
rs147795656 Chr.7: 41700102-41700102 on Build GRCh38
rs762653354 Chr.7: 41700158-41700158 on Build GRCh38
rs149702280 Chr.7: 41700081-41700081 on Build GRCh38

View Full Product Details