Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136430200-136430463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5E
Entrez Gene ID
56623
Gene Name
inositol polyphosphate-5-phosphatase E
Gene Aliases
CORS1, CPD4, JBTS1, MORMS, PPI5PIV
UniGene
Hs.120998
Gene Chromosome Location
Chr.9: 136428615-136439861 on Build GRCh38
Transcripts
NM_001318502.1 NM_019892.5 XM_017014926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220568 c.1747G>A p.D583N Chr.9: 136430332-136430332 on Build GRCh38
4650828 c.1672G>A p.V558I Chr.9: 136430407-136430407 on Build GRCh38
7230414 c.1791G>A p.P597P Chr.9: 136430288-136430288 on Build GRCh38
3367641 c.1696G>T p.G566C Chr.9: 136430383-136430383 on Build GRCh38
5359439 c.1668C>T p.D556D Chr.9: 136430411-136430411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756888841 Chr.9: 136430332-136430332 on Build GRCh38
rs888610698 Chr.9: 136430363-136430363 on Build GRCh38
rs746644044 Chr.9: 136430229-136430229 on Build GRCh38
rs769104210 Chr.9: 136430364-136430364 on Build GRCh38
rs376839080 Chr.9: 136430404-136430404 on Build GRCh38
rs371960390 Chr.9: 136430392-136430392 on Build GRCh38
rs377290486 Chr.9: 136430226-136430226 on Build GRCh38
rs370460379 Chr.9: 136430237-136430237 on Build GRCh38
rs550876114 Chr.9: 136430426-136430426 on Build GRCh38
rs187296291 Chr.9: 136430248-136430248 on Build GRCh38
rs1033746159 Chr.9: 136430258-136430258 on Build GRCh38
rs33982662 Chr.9: 136430285-136430285 on Build GRCh38
rs761647354 Chr.9: 136430439-136430439 on Build GRCh38
rs925789744 Chr.9: 136430305-136430305 on Build GRCh38
rs751316714 Chr.9: 136430331-136430331 on Build GRCh38
rs745341543 Chr.9: 136430262-136430262 on Build GRCh38
rs775518991 Chr.9: 136430319-136430319 on Build GRCh38
rs763992407 Chr.9: 136430326-136430326 on Build GRCh38
rs952748282 Chr.9: 136430233-136430233 on Build GRCh38
rs765327224 Chr.9: 136430292-136430292 on Build GRCh38
rs368026621 Chr.9: 136430336-136430336 on Build GRCh38
rs776792436 Chr.9: 136430408-136430408 on Build GRCh38
rs958733672 Chr.9: 136430253-136430253 on Build GRCh38
rs992814593 Chr.9: 136430409-136430409 on Build GRCh38
rs868448861 Chr.9: 136430227-136430227 on Build GRCh38
rs1025785482 Chr.9: 136430358-136430358 on Build GRCh38
rs369685858 Chr.9: 136430418-136430418 on Build GRCh38
rs943471686 Chr.9: 136430337-136430337 on Build GRCh38
rs121918128 Chr.9: 136430391-136430391 on Build GRCh38
rs750836133 Chr.9: 136430283-136430283 on Build GRCh38
rs368296709 Chr.9: 136430365-136430365 on Build GRCh38
rs778536210 Chr.9: 136430386-136430386 on Build GRCh38
rs559636009 Chr.9: 136430347-136430347 on Build GRCh38
rs1032214000 Chr.9: 136430220-136430220 on Build GRCh38
rs143552175 Chr.9: 136430309-136430309 on Build GRCh38
rs10870182 Chr.9: 136430288-136430288 on Build GRCh38
rs374671289 Chr.9: 136430432-136430432 on Build GRCh38
rs749810093 Chr.9: 136430356-136430356 on Build GRCh38
rs1033384371 Chr.9: 136430443-136430443 on Build GRCh38
rs760729838 Chr.9: 136430393-136430393 on Build GRCh38
rs779177166 Chr.9: 136430379-136430379 on Build GRCh38
rs574429967 Chr.9: 136430225-136430225 on Build GRCh38
rs760790290 Chr.9: 136430289-136430289 on Build GRCh38
rs202207991 Chr.9: 136430234-136430234 on Build GRCh38
rs374679907 Chr.9: 136430263-136430263 on Build GRCh38
rs80111347 Chr.9: 136430397-136430397 on Build GRCh38
rs1024279229 Chr.9: 136430410-136430410 on Build GRCh38
rs775953920 Chr.9: 136430269-136430269 on Build GRCh38
rs372545147 Chr.9: 136430425-136430425 on Build GRCh38
rs749720435 Chr.9: 136430277-136430277 on Build GRCh38
rs1003855913 Chr.9: 136430442-136430442 on Build GRCh38
rs201127434 Chr.9: 136430251-136430251 on Build GRCh38
rs863225197 Chr.9: 136430395-136430395 on Build GRCh38
rs530758804 Chr.9: 136430291-136430291 on Build GRCh38
rs375636136 Chr.9: 136430231-136430231 on Build GRCh38
rs147967974 Chr.9: 136430349-136430349 on Build GRCh38
rs375615380 Chr.9: 136430357-136430357 on Build GRCh38
rs983827023 Chr.9: 136430267-136430267 on Build GRCh38
rs140543689 Chr.9: 136430382-136430382 on Build GRCh38
rs752300607 Chr.9: 136430325-136430325 on Build GRCh38
rs371946549 Chr.9: 136430348-136430348 on Build GRCh38
rs888901624 Chr.9: 136430333-136430333 on Build GRCh38
rs4451431 Chr.9: 136430228-136430228 on Build GRCh38
rs368615705 Chr.9: 136430378-136430378 on Build GRCh38

View Full Product Details