Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131162489-131162762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT1
Entrez Gene ID
3094
Gene Name
histidine triad nucleotide binding protein 1
Gene Aliases
HINT, NMAN, PKCI-1, PRKCNH1
UniGene
Hs.483305
Gene Chromosome Location
Chr.5: 131159283-131165348 on Build GRCh38
Transcripts
NM_005340.6 NR_024610.2 NR_024611.2 NR_073488.1 NR_134494.1 NR_134495.1 XM_017009410.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996483 c.139C>G p.Q47E Chr.5: 131162649-131162649 on Build GRCh38
1060516 c.112-1G>A p.? Chr.5: 131162677-131162677 on Build GRCh38
1060515 c.171G>T p.K57N Chr.5: 131162617-131162617 on Build GRCh38
5984861 c.202G>A p.D68N Chr.5: 131162586-131162586 on Build GRCh38
3669375 c.181T>G p.S61A Chr.5: 131162607-131162607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751105694 Chr.5: 131162563-131162563 on Build GRCh38
rs397514491 Chr.5: 131162636-131162636 on Build GRCh38
rs751481922 Chr.5: 131162689-131162689 on Build GRCh38
rs376643598 Chr.5: 131162688-131162688 on Build GRCh38
rs762701283 Chr.5: 131162676-131162676 on Build GRCh38
rs397514492 Chr.5: 131162604-131162604 on Build GRCh38
rs371722201 Chr.5: 131162594-131162594 on Build GRCh38
rs368167215 Chr.5: 131162640-131162640 on Build GRCh38
rs149224817 Chr.5: 131162536-131162536 on Build GRCh38
rs199716973 Chr.5: 131162629-131162629 on Build GRCh38
rs17165688 Chr.5: 131162535-131162535 on Build GRCh38
rs753183844 Chr.5: 131162727-131162727 on Build GRCh38
rs776900090 Chr.5: 131162529-131162529 on Build GRCh38
rs78949626 Chr.5: 131162671-131162671 on Build GRCh38
rs376264129 Chr.5: 131162596-131162596 on Build GRCh38
rs796918292 Chr.5: 131162535-131162536 on Build GRCh38
rs767015356 Chr.5: 131162702-131162702 on Build GRCh38
rs761323995 Chr.5: 131162701-131162701 on Build GRCh38
rs890477826 Chr.5: 131162603-131162603 on Build GRCh38
rs769497693 Chr.5: 131162670-131162670 on Build GRCh38
rs776720598 Chr.5: 131162660-131162660 on Build GRCh38
rs370640868 Chr.5: 131162620-131162620 on Build GRCh38
rs374733986 Chr.5: 131162601-131162601 on Build GRCh38
rs779545559 Chr.5: 131162721-131162721 on Build GRCh38
rs748504871 Chr.5: 131162599-131162599 on Build GRCh38
rs759762924 Chr.5: 131162665-131162665 on Build GRCh38
rs1048322333 Chr.5: 131162600-131162600 on Build GRCh38
rs763997479 Chr.5: 131162548-131162548 on Build GRCh38
rs750027964 Chr.5: 131162577-131162577 on Build GRCh38
rs369955429 Chr.5: 131162706-131162706 on Build GRCh38
rs1802917 Chr.5: 131162653-131162653 on Build GRCh38
rs371048016 Chr.5: 131162585-131162585 on Build GRCh38
rs143377065 Chr.5: 131162713-131162713 on Build GRCh38
rs199967337 Chr.5: 131162650-131162650 on Build GRCh38
rs758045140 Chr.5: 131162597-131162597 on Build GRCh38
rs780762280 Chr.5: 131162572-131162572 on Build GRCh38
rs866877166 Chr.5: 131162628-131162628 on Build GRCh38
rs150581567 Chr.5: 131162590-131162590 on Build GRCh38

View Full Product Details