Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28787718-28787969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS9
Entrez Gene ID
55756
Gene Name
integrator complex subunit 9
Gene Aliases
CPSF2L, INT9, RC74
UniGene
Hs.162397
Gene Chromosome Location
Chr.8: 28767658-28890226 on Build GRCh38
Transcripts
NM_001145159.2 NM_001172562.1 NM_018250.3 NR_026826.2 XM_006716356.2 XM_006716357.3 XM_011544573.1 XM_011544574.1 XM_011544575.2 XM_011544576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272399 c.1094C>A p.A365E Chr.8: 28787833-28787833 on Build GRCh38
3367307 c.1085T>G p.F362C Chr.8: 28787842-28787842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774139148 Chr.8: 28787839-28787839 on Build GRCh38
rs773661159 Chr.8: 28787872-28787875 on Build GRCh38
rs770036352 Chr.8: 28787926-28787926 on Build GRCh38
rs780652257 Chr.8: 28787901-28787901 on Build GRCh38
rs187513387 Chr.8: 28787755-28787755 on Build GRCh38
rs763630161 Chr.8: 28787934-28787934 on Build GRCh38
rs774283265 Chr.8: 28787902-28787902 on Build GRCh38
rs755712559 Chr.8: 28787871-28787871 on Build GRCh38
rs373334741 Chr.8: 28787933-28787933 on Build GRCh38
rs768621140 Chr.8: 28787815-28787819 on Build GRCh38
rs756335957 Chr.8: 28787798-28787798 on Build GRCh38
rs753483725 Chr.8: 28787880-28787880 on Build GRCh38
rs368666382 Chr.8: 28787929-28787929 on Build GRCh38
rs779379702 Chr.8: 28787892-28787892 on Build GRCh38
rs763636831 Chr.8: 28787878-28787878 on Build GRCh38
rs200868611 Chr.8: 28787895-28787895 on Build GRCh38
rs775551761 Chr.8: 28787932-28787932 on Build GRCh38
rs1006542056 Chr.8: 28787775-28787775 on Build GRCh38
rs767348752 Chr.8: 28787931-28787931 on Build GRCh38
rs780211763 Chr.8: 28787918-28787918 on Build GRCh38
rs765033027 Chr.8: 28787939-28787939 on Build GRCh38
rs372014318 Chr.8: 28787845-28787845 on Build GRCh38
rs752113604 Chr.8: 28787860-28787860 on Build GRCh38
rs774108152 Chr.8: 28787927-28787927 on Build GRCh38
rs768842925 Chr.8: 28787904-28787904 on Build GRCh38
rs953720717 Chr.8: 28787774-28787774 on Build GRCh38
rs241187 Chr.8: 28787754-28787754 on Build GRCh38
rs376857624 Chr.8: 28787896-28787896 on Build GRCh38
rs756918843 Chr.8: 28787938-28787938 on Build GRCh38
rs112863183 Chr.8: 28787908-28787908 on Build GRCh38
rs747055117 Chr.8: 28787903-28787903 on Build GRCh38
rs749395089 Chr.8: 28787925-28787925 on Build GRCh38
rs762727736 Chr.8: 28787809-28787809 on Build GRCh38
rs756973998 Chr.8: 28787891-28787891 on Build GRCh38
rs766107755 Chr.8: 28787835-28787835 on Build GRCh38
rs767536500 Chr.8: 28787855-28787855 on Build GRCh38
rs566056873 Chr.8: 28787773-28787773 on Build GRCh38

View Full Product Details