Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50318028-50318275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL2
Entrez Gene ID
8692
Gene Name
hyaluronoglucosaminidase 2
Gene Aliases
LUCA2
UniGene
Hs.76873
Gene Chromosome Location
Chr.3: 50317790-50322850 on Build GRCh38
Transcripts
NM_003773.4 NM_033158.4 XM_005265524.2 XM_005265525.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1424161 c.1394C>T p.A465V Chr.3: 50318157-50318157 on Build GRCh38
5834701 c.1301_1302insAA p.S434fs*>41 Chr.3: 50318249-50318250 on Build GRCh38
7186631 c.1357T>A p.W453R Chr.3: 50318194-50318194 on Build GRCh38
1670216 c.1304G>A p.G435D Chr.3: 50318247-50318247 on Build GRCh38
5834700 c.1307delA p.E436fs*>38 Chr.3: 50318244-50318244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587730990 Chr.3: 50318225-50318225 on Build GRCh38
rs374329099 Chr.3: 50318154-50318154 on Build GRCh38
rs782118213 Chr.3: 50318240-50318240 on Build GRCh38
rs782404084 Chr.3: 50318106-50318106 on Build GRCh38
rs782005378 Chr.3: 50318235-50318235 on Build GRCh38
rs782710947 Chr.3: 50318127-50318127 on Build GRCh38
rs782428726 Chr.3: 50318174-50318174 on Build GRCh38
rs954448508 Chr.3: 50318072-50318072 on Build GRCh38
rs75801519 Chr.3: 50318173-50318173 on Build GRCh38
rs142551472 Chr.3: 50318150-50318150 on Build GRCh38
rs782779110 Chr.3: 50318141-50318141 on Build GRCh38
rs782404838 Chr.3: 50318233-50318233 on Build GRCh38
rs587620868 Chr.3: 50318226-50318226 on Build GRCh38
rs587606595 Chr.3: 50318067-50318067 on Build GRCh38
rs373307469 Chr.3: 50318200-50318200 on Build GRCh38
rs781931505 Chr.3: 50318208-50318208 on Build GRCh38
rs782339100 Chr.3: 50318202-50318202 on Build GRCh38
rs782599975 Chr.3: 50318169-50318169 on Build GRCh38
rs1021534005 Chr.3: 50318054-50318054 on Build GRCh38
rs781907096 Chr.3: 50318137-50318137 on Build GRCh38
rs782010090 Chr.3: 50318085-50318085 on Build GRCh38
rs782077725 Chr.3: 50318121-50318121 on Build GRCh38
rs782133278 Chr.3: 50318094-50318094 on Build GRCh38
rs200925068 Chr.3: 50318080-50318080 on Build GRCh38
rs782228571 Chr.3: 50318201-50318201 on Build GRCh38
rs782167270 Chr.3: 50318055-50318055 on Build GRCh38
rs782685136 Chr.3: 50318175-50318175 on Build GRCh38
rs782152752 Chr.3: 50318138-50318138 on Build GRCh38
rs782272709 Chr.3: 50318192-50318192 on Build GRCh38
rs920815254 Chr.3: 50318132-50318132 on Build GRCh38
rs1050139 Chr.3: 50318078-50318078 on Build GRCh38
rs80031801 Chr.3: 50318117-50318117 on Build GRCh38
rs771280335 Chr.3: 50318083-50318083 on Build GRCh38

View Full Product Details