Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156624224-156624495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN2
Entrez Gene ID
60484
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
BRAL1
UniGene
Hs.410719
Gene Chromosome Location
Chr.1: 156601504-156626608 on Build GRCh38
Transcripts
NM_021817.2 XM_005245415.2 XM_011509853.2 XM_017002020.1 XM_017002021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1578804 c.473G>A p.R158Q Chr.1: 156624384-156624384 on Build GRCh38
6229629 c.440-5C>A p.? Chr.1: 156624346-156624346 on Build GRCh38
1719028 c.451C>T p.P151S Chr.1: 156624362-156624362 on Build GRCh38
897277 c.440-2A>G p.? Chr.1: 156624349-156624349 on Build GRCh38
4023344 c.487T>C p.Y163H Chr.1: 156624398-156624398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755747077 Chr.1: 156624449-156624449 on Build GRCh38
rs776749289 Chr.1: 156624346-156624346 on Build GRCh38
rs376296474 Chr.1: 156624327-156624327 on Build GRCh38
rs762072620 Chr.1: 156624353-156624353 on Build GRCh38
rs750476459 Chr.1: 156624362-156624362 on Build GRCh38
rs781729246 Chr.1: 156624304-156624304 on Build GRCh38
rs766936737 Chr.1: 156624379-156624379 on Build GRCh38
rs770331311 Chr.1: 156624474-156624474 on Build GRCh38
rs747800991 Chr.1: 156624306-156624306 on Build GRCh38
rs367871357 Chr.1: 156624299-156624299 on Build GRCh38
rs769375024 Chr.1: 156624317-156624317 on Build GRCh38
rs770830866 Chr.1: 156624331-156624331 on Build GRCh38
rs759891872 Chr.1: 156624371-156624371 on Build GRCh38
rs553670177 Chr.1: 156624367-156624367 on Build GRCh38
rs765399919 Chr.1: 156624354-156624354 on Build GRCh38
rs148744711 Chr.1: 156624466-156624466 on Build GRCh38
rs926447540 Chr.1: 156624329-156624329 on Build GRCh38
rs755408840 Chr.1: 156624302-156624302 on Build GRCh38
rs955404462 Chr.1: 156624279-156624279 on Build GRCh38
rs762947888 Chr.1: 156624373-156624373 on Build GRCh38
rs774282053 Chr.1: 156624335-156624335 on Build GRCh38
rs369574575 Chr.1: 156624340-156624340 on Build GRCh38
rs778458667 Chr.1: 156624477-156624477 on Build GRCh38
rs1030136819 Chr.1: 156624256-156624256 on Build GRCh38
rs959441276 Chr.1: 156624343-156624343 on Build GRCh38
rs372076575 Chr.1: 156624318-156624318 on Build GRCh38
rs886978074 Chr.1: 156624454-156624454 on Build GRCh38
rs753055672 Chr.1: 156624441-156624441 on Build GRCh38
rs752100697 Chr.1: 156624398-156624398 on Build GRCh38
rs777291324 Chr.1: 156624453-156624453 on Build GRCh38
rs566088886 Chr.1: 156624417-156624417 on Build GRCh38
rs981191927 Chr.1: 156624312-156624312 on Build GRCh38
rs898421912 Chr.1: 156624328-156624328 on Build GRCh38
rs781668667 Chr.1: 156624436-156624436 on Build GRCh38
rs770028647 Chr.1: 156624405-156624405 on Build GRCh38
rs928994323 Chr.1: 156624358-156624358 on Build GRCh38

View Full Product Details