Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22120487-22120750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
UniGene
Hs.272367
Gene Chromosome Location
Chr.8: 22114419-22131053 on Build GRCh38
Transcripts
NM_005144.4 NM_018411.4 XM_005273569.1 XM_006716367.1

SNP Detail

SNP ID Chromosome Location
rs968023413 Chr.8: 22120663-22120663 on Build GRCh38
rs909542980 Chr.8: 22120587-22120587 on Build GRCh38
rs755993112 Chr.8: 22120690-22120694 on Build GRCh38
rs1041391347 Chr.8: 22120683-22120683 on Build GRCh38
rs889996237 Chr.8: 22120635-22120635 on Build GRCh38
rs1031063452 Chr.8: 22120680-22120680 on Build GRCh38
rs989831319 Chr.8: 22120654-22120654 on Build GRCh38
rs766806568 Chr.8: 22120690-22120690 on Build GRCh38
rs1051289987 Chr.8: 22120622-22120622 on Build GRCh38
rs1055743626 Chr.8: 22120727-22120727 on Build GRCh38
rs1044420907 Chr.8: 22120586-22120586 on Build GRCh38
rs755280580 Chr.8: 22120533-22120533 on Build GRCh38
rs770377560 Chr.8: 22120555-22120555 on Build GRCh38
rs781194153 Chr.8: 22120534-22120536 on Build GRCh38
rs751722289 Chr.8: 22120530-22120530 on Build GRCh38
rs761322319 Chr.8: 22120688-22120688 on Build GRCh38
rs1007149880 Chr.8: 22120643-22120643 on Build GRCh38
rs774827634 Chr.8: 22120558-22120558 on Build GRCh38
rs558642481 Chr.8: 22120601-22120601 on Build GRCh38
rs941067816 Chr.8: 22120600-22120600 on Build GRCh38
rs575060011 Chr.8: 22120701-22120701 on Build GRCh38
rs77452325 Chr.8: 22120689-22120689 on Build GRCh38
rs948669412 Chr.8: 22120585-22120585 on Build GRCh38
rs376800273 Chr.8: 22120515-22120515 on Build GRCh38
rs765241716 Chr.8: 22120561-22120561 on Build GRCh38
rs1026275848 Chr.8: 22120569-22120569 on Build GRCh38
rs749599102 Chr.8: 22120556-22120556 on Build GRCh38
rs779962253 Chr.8: 22120522-22120522 on Build GRCh38
rs117242897 Chr.8: 22120511-22120511 on Build GRCh38
rs1017047365 Chr.8: 22120528-22120528 on Build GRCh38
rs768383276 Chr.8: 22120667-22120667 on Build GRCh38
rs745732994 Chr.8: 22120671-22120671 on Build GRCh38
rs924047060 Chr.8: 22120532-22120532 on Build GRCh38
rs1023003084 Chr.8: 22120655-22120655 on Build GRCh38
rs955655650 Chr.8: 22120574-22120574 on Build GRCh38
rs763121161 Chr.8: 22120673-22120673 on Build GRCh38
rs188408063 Chr.8: 22120591-22120591 on Build GRCh38
rs577013634 Chr.8: 22120603-22120603 on Build GRCh38
rs776068377 Chr.8: 22120677-22120677 on Build GRCh38
rs781105215 Chr.8: 22120547-22120547 on Build GRCh38
rs11135859 Chr.8: 22120559-22120559 on Build GRCh38
rs750351919 Chr.8: 22120699-22120699 on Build GRCh38
rs998277491 Chr.8: 22120620-22120620 on Build GRCh38
rs11779694 Chr.8: 22120575-22120575 on Build GRCh38
rs750157785 Chr.8: 22120513-22120513 on Build GRCh38
rs748271794 Chr.8: 22120548-22120548 on Build GRCh38
rs544189582 Chr.8: 22120666-22120666 on Build GRCh38

View Full Product Details