Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8867647-8867896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMX1
Entrez Gene ID
3166
Gene Name
H6 family homeobox 1
Gene Aliases
H6, NKX5-3
UniGene
Hs.104134
Gene Chromosome Location
Chr.4: 8846076-8871817 on Build GRCh38
Transcripts
NM_001306142.1 NM_018942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377778 c.1021C>T p.R341W Chr.4: 8867719-8867719 on Build GRCh38
6545269 c.1002C>A p.A334A Chr.4: 8867738-8867738 on Build GRCh38
1619201 c.879A>T p.A293A Chr.4: 8867861-8867861 on Build GRCh38
1659225 c.866G>A p.S289N Chr.4: 8867874-8867874 on Build GRCh38
7276507 c.1000G>A p.A334T Chr.4: 8867740-8867740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763505877 Chr.4: 8867776-8867776 on Build GRCh38
rs757578757 Chr.4: 8867834-8867834 on Build GRCh38
rs776532962 Chr.4: 8867862-8867862 on Build GRCh38
rs1020239422 Chr.4: 8867880-8867880 on Build GRCh38
rs901630227 Chr.4: 8867827-8867827 on Build GRCh38
rs930860616 Chr.4: 8867740-8867740 on Build GRCh38
rs940379768 Chr.4: 8867799-8867799 on Build GRCh38
rs1050213588 Chr.4: 8867842-8867842 on Build GRCh38
rs775671619 Chr.4: 8867873-8867873 on Build GRCh38
rs987867878 Chr.4: 8867692-8867692 on Build GRCh38
rs907520239 Chr.4: 8867777-8867777 on Build GRCh38
rs752877327 Chr.4: 8867739-8867739 on Build GRCh38
rs746059506 Chr.4: 8867868-8867868 on Build GRCh38
rs778237993 Chr.4: 8867849-8867849 on Build GRCh38
rs1037767196 Chr.4: 8867728-8867728 on Build GRCh38
rs960901073 Chr.4: 8867669-8867669 on Build GRCh38
rs747004731 Chr.4: 8867858-8867858 on Build GRCh38
rs943428959 Chr.4: 8867716-8867716 on Build GRCh38
rs569192304 Chr.4: 8867741-8867741 on Build GRCh38
rs759871570 Chr.4: 8867713-8867713 on Build GRCh38
rs910580880 Chr.4: 8867702-8867702 on Build GRCh38
rs765203669 Chr.4: 8867714-8867714 on Build GRCh38
rs550799914 Chr.4: 8867715-8867715 on Build GRCh38
rs754707432 Chr.4: 8867848-8867848 on Build GRCh38
rs753400835 Chr.4: 8867847-8867847 on Build GRCh38
rs539490551 Chr.4: 8867844-8867844 on Build GRCh38
rs933276759 Chr.4: 8867691-8867691 on Build GRCh38
rs770080827 Chr.4: 8867872-8867872 on Build GRCh38
rs758262483 Chr.4: 8867853-8867853 on Build GRCh38
rs1040106231 Chr.4: 8867818-8867818 on Build GRCh38
rs751908261 Chr.4: 8867796-8867796 on Build GRCh38
rs747683664 Chr.4: 8867852-8867852 on Build GRCh38
rs764730446 Chr.4: 8867785-8867785 on Build GRCh38
rs13303225 Chr.4: 8867861-8867861 on Build GRCh38
rs1011420309 Chr.4: 8867840-8867863 on Build GRCh38
rs777697331 Chr.4: 8867854-8867854 on Build GRCh38
rs13302936 Chr.4: 8867841-8867841 on Build GRCh38
rs535529989 Chr.4: 8867704-8867704 on Build GRCh38

View Full Product Details