Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74529889-74530140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUP1
Entrez Gene ID
550
Gene Name
ancient ubiquitous protein 1
Gene Aliases
-
UniGene
Hs.411480
Gene Chromosome Location
Chr.2: 74526648-74529897 on Build GRCh38
Transcripts
NM_181575.4 NR_126510.1 NR_126511.1 NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Gene Symbol
HTRA2
Entrez Gene ID
27429
Gene Name
HtrA serine peptidase 2
Gene Aliases
OMI, PARK13, PRSS25
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74529405-74533556 on Build GRCh38
Transcripts
NM_181575.4 NR_126510.1 NR_126511.1 NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166160 c.43C>T p.R15W Chr.2: 74530049-74530049 on Build GRCh38
4315338 c.18G>A p.A6A Chr.2: 74530024-74530024 on Build GRCh38
4141251 c.45G>C p.R15R Chr.2: 74530051-74530051 on Build GRCh38
419101 c.12G>A p.P4P Chr.2: 74530018-74530018 on Build GRCh38
6683081 c.5C>A p.A2D Chr.2: 74530011-74530011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032522339 Chr.2: 74529915-74529915 on Build GRCh38
rs768870295 Chr.2: 74530068-74530068 on Build GRCh38
rs778811054 Chr.2: 74530023-74530023 on Build GRCh38
rs866473711 Chr.2: 74530058-74530058 on Build GRCh38
rs749586893 Chr.2: 74530045-74530045 on Build GRCh38
rs1019901895 Chr.2: 74529951-74529951 on Build GRCh38
rs749968474 Chr.2: 74529990-74529990 on Build GRCh38
rs917910498 Chr.2: 74530052-74530052 on Build GRCh38
rs764609801 Chr.2: 74529967-74529967 on Build GRCh38
rs774713939 Chr.2: 74529971-74529971 on Build GRCh38
rs886056344 Chr.2: 74529921-74529921 on Build GRCh38
rs772527738 Chr.2: 74530102-74530102 on Build GRCh38
rs762421882 Chr.2: 74529976-74529976 on Build GRCh38
rs781093029 Chr.2: 74530029-74530029 on Build GRCh38
rs949523442 Chr.2: 74530082-74530082 on Build GRCh38
rs775932862 Chr.2: 74529964-74529964 on Build GRCh38
rs995598480 Chr.2: 74530051-74530051 on Build GRCh38
rs765943892 Chr.2: 74530112-74530112 on Build GRCh38
rs965436814 Chr.2: 74529942-74529942 on Build GRCh38
rs955155115 Chr.2: 74529959-74529959 on Build GRCh38
rs374912838 Chr.2: 74529960-74529960 on Build GRCh38
rs1019900133 Chr.2: 74530074-74530074 on Build GRCh38
rs963881310 Chr.2: 74529983-74529983 on Build GRCh38
rs770085229 Chr.2: 74530036-74530036 on Build GRCh38
rs753568807 Chr.2: 74530016-74530016 on Build GRCh38
rs768096748 Chr.2: 74530093-74530093 on Build GRCh38
rs957038788 Chr.2: 74529921-74529921 on Build GRCh38
rs368984550 Chr.2: 74529963-74529963 on Build GRCh38
rs770914677 Chr.2: 74530111-74530111 on Build GRCh38
rs372441631 Chr.2: 74530024-74530024 on Build GRCh38
rs745948813 Chr.2: 74530032-74530032 on Build GRCh38
rs766021850 Chr.2: 74530015-74530015 on Build GRCh38
rs775840965 Chr.2: 74530042-74530042 on Build GRCh38
rs754766915 Chr.2: 74530022-74530022 on Build GRCh38
rs547013664 Chr.2: 74529917-74529917 on Build GRCh38
rs760187812 Chr.2: 74530108-74530108 on Build GRCh38
rs2231249 Chr.2: 74530014-74530014 on Build GRCh38
rs766913545 Chr.2: 74529985-74529985 on Build GRCh38
rs913792802 Chr.2: 74529965-74529965 on Build GRCh38
rs774827622 Chr.2: 74530071-74530071 on Build GRCh38
rs145946172 Chr.2: 74530083-74530083 on Build GRCh38
rs749132601 Chr.2: 74529943-74529943 on Build GRCh38
rs945306746 Chr.2: 74529969-74529969 on Build GRCh38
rs557990839 Chr.2: 74530119-74530119 on Build GRCh38
rs539659478 Chr.2: 74529918-74529918 on Build GRCh38

View Full Product Details