Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82196255-82196522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IBTK
Entrez Gene ID
25998
Gene Name
inhibitor of Bruton tyrosine kinase
Gene Aliases
BTBD26, BTKI
UniGene
Hs.306425
Gene Chromosome Location
Chr.6: 82169987-82248294 on Build GRCh38
Transcripts
NM_001300906.1 NM_015525.3 XM_006715453.2 XM_006715454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4438851 c.3064C>T p.L1022F Chr.6: 82196408-82196408 on Build GRCh38
4438850 c.3157C>T p.H1053Y Chr.6: 82196315-82196315 on Build GRCh38
1081966 c.3114T>C p.G1038G Chr.6: 82196358-82196358 on Build GRCh38
7351823 c.3026-2A>C p.? Chr.6: 82196448-82196448 on Build GRCh38
6529871 c.3086A>G p.D1029G Chr.6: 82196386-82196386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768793874 Chr.6: 82196306-82196306 on Build GRCh38
rs774472425 Chr.6: 82196462-82196463 on Build GRCh38
rs748196965 Chr.6: 82196482-82196482 on Build GRCh38
rs778674707 Chr.6: 82196439-82196439 on Build GRCh38
rs907243153 Chr.6: 82196297-82196297 on Build GRCh38
rs536608941 Chr.6: 82196483-82196483 on Build GRCh38
rs757016766 Chr.6: 82196427-82196427 on Build GRCh38
rs374259831 Chr.6: 82196310-82196310 on Build GRCh38
rs769029721 Chr.6: 82196472-82196472 on Build GRCh38
rs769826293 Chr.6: 82196490-82196490 on Build GRCh38
rs753270442 Chr.6: 82196422-82196422 on Build GRCh38
rs1021524276 Chr.6: 82196462-82196462 on Build GRCh38
rs770057275 Chr.6: 82196358-82196359 on Build GRCh38
rs1047062116 Chr.6: 82196436-82196436 on Build GRCh38
rs759360099 Chr.6: 82196365-82196365 on Build GRCh38
rs919854666 Chr.6: 82196295-82196295 on Build GRCh38
rs138739466 Chr.6: 82196481-82196481 on Build GRCh38
rs765425757 Chr.6: 82196323-82196323 on Build GRCh38
rs867416502 Chr.6: 82196309-82196309 on Build GRCh38
rs1011520289 Chr.6: 82196455-82196455 on Build GRCh38
rs113751560 Chr.6: 82196396-82196396 on Build GRCh38
rs780224469 Chr.6: 82196462-82196462 on Build GRCh38
rs965148923 Chr.6: 82196485-82196485 on Build GRCh38
rs188420300 Chr.6: 82196446-82196446 on Build GRCh38
rs773448650 Chr.6: 82196494-82196494 on Build GRCh38
rs775629198 Chr.6: 82196364-82196375 on Build GRCh38
rs1031661560 Chr.6: 82196340-82196340 on Build GRCh38
rs752696580 Chr.6: 82196391-82196391 on Build GRCh38
rs538617195 Chr.6: 82196471-82196471 on Build GRCh38
rs777405062 Chr.6: 82196420-82196420 on Build GRCh38
rs751397840 Chr.6: 82196349-82196349 on Build GRCh38
rs764036027 Chr.6: 82196461-82196462 on Build GRCh38
rs377724789 Chr.6: 82196368-82196368 on Build GRCh38
rs369544247 Chr.6: 82196298-82196298 on Build GRCh38
rs747554888 Chr.6: 82196470-82196470 on Build GRCh38
rs762153857 Chr.6: 82196316-82196316 on Build GRCh38
rs762271271 Chr.6: 82196478-82196492 on Build GRCh38
rs772585649 Chr.6: 82196447-82196447 on Build GRCh38
rs534851417 Chr.6: 82196397-82196397 on Build GRCh38
rs377219714 Chr.6: 82196283-82196283 on Build GRCh38
rs763020878 Chr.6: 82196392-82196393 on Build GRCh38

View Full Product Details