Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62347057-62347284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855796 c.5T>C p.V2A Chr.5: 62347151-62347151 on Build GRCh38
4616129 c.154_155delAA p.K52fs*3 Chr.5: 62347219-62347220 on Build GRCh38
1212451 c.100G>T p.E34* Chr.5: 62347165-62347165 on Build GRCh38
3617328 c.11C>T p.S4F Chr.5: 62347157-62347157 on Build GRCh38
3855795 c.86T>C p.V29A Chr.5: 62347151-62347151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039433409 Chr.5: 62347224-62347224 on Build GRCh38
rs899555129 Chr.5: 62347240-62347240 on Build GRCh38
rs753309327 Chr.5: 62347113-62347113 on Build GRCh38
rs745977169 Chr.5: 62347251-62347251 on Build GRCh38
rs751850670 Chr.5: 62347178-62347178 on Build GRCh38
rs925638530 Chr.5: 62347172-62347172 on Build GRCh38
rs917541316 Chr.5: 62347086-62347086 on Build GRCh38
rs765160141 Chr.5: 62347154-62347154 on Build GRCh38
rs150192070 Chr.5: 62347168-62347168 on Build GRCh38
rs746125264 Chr.5: 62347247-62347248 on Build GRCh38
rs768845624 Chr.5: 62347103-62347103 on Build GRCh38
rs776102613 Chr.5: 62347085-62347085 on Build GRCh38
rs757402774 Chr.5: 62347212-62347212 on Build GRCh38
rs754330795 Chr.5: 62347090-62347093 on Build GRCh38
rs759003955 Chr.5: 62347126-62347126 on Build GRCh38
rs922141597 Chr.5: 62347180-62347180 on Build GRCh38
rs546664729 Chr.5: 62347175-62347175 on Build GRCh38
rs1032468118 Chr.5: 62347115-62347115 on Build GRCh38
rs752570320 Chr.5: 62347167-62347167 on Build GRCh38
rs763414339 Chr.5: 62347099-62347099 on Build GRCh38
rs779321122 Chr.5: 62347229-62347230 on Build GRCh38
rs757760627 Chr.5: 62347226-62347227 on Build GRCh38
rs3213940 Chr.5: 62347110-62347110 on Build GRCh38
rs772983615 Chr.5: 62347108-62347108 on Build GRCh38
rs760213648 Chr.5: 62347109-62347109 on Build GRCh38
rs966360584 Chr.5: 62347111-62347111 on Build GRCh38
rs1013492020 Chr.5: 62347107-62347107 on Build GRCh38
rs781430441 Chr.5: 62347250-62347250 on Build GRCh38

View Full Product Details