Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41724518-41724777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAP1
Entrez Gene ID
9001
Gene Name
huntingtin associated protein 1
Gene Aliases
HAP2, HIP5, HLP, hHLP1
UniGene
Hs.158300
Gene Chromosome Location
Chr.17: 41722639-41734646 on Build GRCh38
Transcripts
NM_001079870.1 NM_001079871.1 NM_177977.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4573241 c.1983_1984CA>AC p.S662R Chr.17: 41724733-41724734 on Build GRCh38
6677561 c.1813delG p.A605fs*>15 Chr.17: 41724748-41724748 on Build GRCh38
7315009 c.1788C>A p.S596R Chr.17: 41724722-41724722 on Build GRCh38
3771357 c.1856G>A p.R619Q Chr.17: 41724705-41724705 on Build GRCh38
7315346 c.1771G>T p.A591S Chr.17: 41724739-41724739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781977398 Chr.17: 41724673-41724673 on Build GRCh38
rs782319651 Chr.17: 41724660-41724660 on Build GRCh38
rs782453513 Chr.17: 41724603-41724603 on Build GRCh38
rs41314142 Chr.17: 41724651-41724651 on Build GRCh38
rs531748071 Chr.17: 41724713-41724713 on Build GRCh38
rs782311151 Chr.17: 41724736-41724736 on Build GRCh38
rs141557284 Chr.17: 41724699-41724699 on Build GRCh38
rs369869133 Chr.17: 41724672-41724672 on Build GRCh38
rs144352175 Chr.17: 41724575-41724575 on Build GRCh38
rs1019093562 Chr.17: 41724630-41724630 on Build GRCh38
rs567948097 Chr.17: 41724580-41724581 on Build GRCh38
rs782590381 Chr.17: 41724662-41724662 on Build GRCh38
rs577955715 Chr.17: 41724642-41724642 on Build GRCh38
rs930481335 Chr.17: 41724683-41724683 on Build GRCh38
rs34044330 Chr.17: 41724751-41724751 on Build GRCh38
rs376429973 Chr.17: 41724676-41724676 on Build GRCh38
rs782082996 Chr.17: 41724754-41724754 on Build GRCh38
rs782692038 Chr.17: 41724753-41724754 on Build GRCh38
rs376345209 Chr.17: 41724669-41724669 on Build GRCh38
rs560765169 Chr.17: 41724653-41724653 on Build GRCh38
rs147264849 Chr.17: 41724729-41724729 on Build GRCh38
rs573325595 Chr.17: 41724600-41724600 on Build GRCh38
rs575840047 Chr.17: 41724581-41724582 on Build GRCh38
rs1020548896 Chr.17: 41724581-41724581 on Build GRCh38
rs782521106 Chr.17: 41724705-41724705 on Build GRCh38
rs781881693 Chr.17: 41724576-41724576 on Build GRCh38
rs200875679 Chr.17: 41724730-41724730 on Build GRCh38
rs190549956 Chr.17: 41724615-41724615 on Build GRCh38
rs199816051 Chr.17: 41724752-41724752 on Build GRCh38
rs376051421 Chr.17: 41724737-41724737 on Build GRCh38
rs201314183 Chr.17: 41724735-41724735 on Build GRCh38
rs782619598 Chr.17: 41724686-41724687 on Build GRCh38
rs781965308 Chr.17: 41724679-41724679 on Build GRCh38
rs782737816 Chr.17: 41724755-41724755 on Build GRCh38
rs34381648 Chr.17: 41724714-41724714 on Build GRCh38
rs965851495 Chr.17: 41724646-41724646 on Build GRCh38
rs782206368 Chr.17: 41724692-41724693 on Build GRCh38
rs78148581 Chr.17: 41724608-41724608 on Build GRCh38
rs369507142 Chr.17: 41724687-41724687 on Build GRCh38
rs199567477 Chr.17: 41724706-41724706 on Build GRCh38
rs782276515 Chr.17: 41724744-41724744 on Build GRCh38
rs148503989 Chr.17: 41724711-41724711 on Build GRCh38
rs374305699 Chr.17: 41724722-41724722 on Build GRCh38
rs782231762 Chr.17: 41724703-41724703 on Build GRCh38
rs966340921 Chr.17: 41724596-41724596 on Build GRCh38

View Full Product Details