Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64376686-64376957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0101
Entrez Gene ID
9768
Gene Name
KIAA0101
Gene Aliases
L5, NS5ATP9, OEATC, OEATC-1, OEATC1, PAF, PAF15, p15(PAF), p15/PAF, p15PAF
UniGene
Hs.81892
Gene Chromosome Location
Chr.15: 64364994-64387687 on Build GRCh38
Transcripts
NM_001029989.2 NM_014736.5 NR_109934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700718 c.167G>C p.R56P Chr.15: 64376866-64376866 on Build GRCh38
963931 c.287G>T p.R96I Chr.15: 64376746-64376746 on Build GRCh38
1301323 c.204C>G p.F68L Chr.15: 64376829-64376829 on Build GRCh38
123896 c.275G>A p.G92E Chr.15: 64376758-64376758 on Build GRCh38
7115918 c.237G>A p.E79E Chr.15: 64376796-64376796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757112395 Chr.15: 64376783-64376783 on Build GRCh38
rs761000759 Chr.15: 64376765-64376765 on Build GRCh38
rs138160608 Chr.15: 64376809-64376809 on Build GRCh38
rs753691125 Chr.15: 64376781-64376781 on Build GRCh38
rs11554320 Chr.15: 64376833-64376833 on Build GRCh38
rs771497961 Chr.15: 64376717-64376717 on Build GRCh38
rs150092906 Chr.15: 64376870-64376870 on Build GRCh38
rs994677332 Chr.15: 64376854-64376854 on Build GRCh38
rs1036414910 Chr.15: 64376780-64376780 on Build GRCh38
rs775654036 Chr.15: 64376895-64376895 on Build GRCh38
rs754634033 Chr.15: 64376853-64376853 on Build GRCh38
rs11554313 Chr.15: 64376798-64376798 on Build GRCh38
rs779642598 Chr.15: 64376829-64376829 on Build GRCh38
rs368370868 Chr.15: 64376921-64376921 on Build GRCh38
rs1007409399 Chr.15: 64376885-64376885 on Build GRCh38
rs750349050 Chr.15: 64376816-64376816 on Build GRCh38
rs867996009 Chr.15: 64376905-64376905 on Build GRCh38
rs11554309 Chr.15: 64376830-64376830 on Build GRCh38
rs886575798 Chr.15: 64376729-64376729 on Build GRCh38
rs767605797 Chr.15: 64376739-64376739 on Build GRCh38
rs140686158 Chr.15: 64376867-64376867 on Build GRCh38
rs774911661 Chr.15: 64376718-64376718 on Build GRCh38
rs549323404 Chr.15: 64376877-64376877 on Build GRCh38
rs780822345 Chr.15: 64376864-64376864 on Build GRCh38
rs774762712 Chr.15: 64376871-64376871 on Build GRCh38
rs375528974 Chr.15: 64376917-64376917 on Build GRCh38
rs760769298 Chr.15: 64376915-64376915 on Build GRCh38
rs757883774 Chr.15: 64376818-64376818 on Build GRCh38
rs374369229 Chr.15: 64376766-64376766 on Build GRCh38
rs1019156695 Chr.15: 64376777-64376777 on Build GRCh38
rs867644942 Chr.15: 64376792-64376792 on Build GRCh38
rs563439020 Chr.15: 64376720-64376720 on Build GRCh38
rs367901741 Chr.15: 64376843-64376843 on Build GRCh38
rs746420273 Chr.15: 64376876-64376876 on Build GRCh38
rs752679515 Chr.15: 64376746-64376746 on Build GRCh38

View Full Product Details