Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14046091-14046362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL27RA
Entrez Gene ID
9466
Gene Name
interleukin 27 receptor subunit alpha
Gene Aliases
CRL1, IL-27RA, IL27R, TCCR, WSX1, zcytor1
UniGene
Hs.132781
Gene Chromosome Location
Chr.19: 14031740-14052919 on Build GRCh38
Transcripts
NM_004843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
474228 c.849C>G p.T283T Chr.19: 14046234-14046234 on Build GRCh38
3529293 c.921G>A p.E307E Chr.19: 14046306-14046306 on Build GRCh38
3362695 c.835C>T p.P279S Chr.19: 14046220-14046220 on Build GRCh38
349508 c.938C>G p.S313C Chr.19: 14046323-14046323 on Build GRCh38
5691952 c.774A>T p.P258P Chr.19: 14046159-14046159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768827448 Chr.19: 14046136-14046136 on Build GRCh38
rs968464483 Chr.19: 14046222-14046222 on Build GRCh38
rs763571022 Chr.19: 14046179-14046179 on Build GRCh38
rs769491247 Chr.19: 14046129-14046129 on Build GRCh38
rs774624578 Chr.19: 14046140-14046140 on Build GRCh38
rs868826157 Chr.19: 14046233-14046233 on Build GRCh38
rs766671315 Chr.19: 14046158-14046158 on Build GRCh38
rs992983922 Chr.19: 14046341-14046341 on Build GRCh38
rs766966794 Chr.19: 14046308-14046308 on Build GRCh38
rs761038516 Chr.19: 14046154-14046154 on Build GRCh38
rs753207679 Chr.19: 14046295-14046295 on Build GRCh38
rs372221939 Chr.19: 14046208-14046208 on Build GRCh38
rs754327642 Chr.19: 14046164-14046164 on Build GRCh38
rs755899251 Chr.19: 14046213-14046213 on Build GRCh38
rs764318693 Chr.19: 14046170-14046170 on Build GRCh38
rs751126050 Chr.19: 14046187-14046187 on Build GRCh38
rs745741708 Chr.19: 14046125-14046125 on Build GRCh38
rs531955454 Chr.19: 14046191-14046191 on Build GRCh38
rs772062615 Chr.19: 14046263-14046263 on Build GRCh38
rs748177405 Chr.19: 14046262-14046262 on Build GRCh38
rs761264810 Chr.19: 14046299-14046299 on Build GRCh38
rs201300850 Chr.19: 14046209-14046209 on Build GRCh38
rs755772171 Chr.19: 14046318-14046318 on Build GRCh38
rs750061223 Chr.19: 14046312-14046312 on Build GRCh38
rs760022121 Chr.19: 14046284-14046284 on Build GRCh38
rs771294505 Chr.19: 14046282-14046282 on Build GRCh38
rs368378512 Chr.19: 14046229-14046229 on Build GRCh38
rs768774659 Chr.19: 14046260-14046260 on Build GRCh38
rs773314639 Chr.19: 14046149-14046149 on Build GRCh38
rs900626558 Chr.19: 14046139-14046139 on Build GRCh38
rs144511816 Chr.19: 14046134-14046134 on Build GRCh38
rs746640333 Chr.19: 14046341-14046342 on Build GRCh38
rs749216683 Chr.19: 14046255-14046255 on Build GRCh38
rs958764923 Chr.19: 14046292-14046292 on Build GRCh38
rs762116471 Chr.19: 14046141-14046141 on Build GRCh38
rs767851552 Chr.19: 14046144-14046144 on Build GRCh38
rs777027035 Chr.19: 14046283-14046283 on Build GRCh38
rs557005103 Chr.19: 14046121-14046121 on Build GRCh38
rs267605301 Chr.19: 14046273-14046273 on Build GRCh38
rs867694672 Chr.19: 14046226-14046226 on Build GRCh38
rs765798453 Chr.19: 14046291-14046291 on Build GRCh38
rs775449661 Chr.19: 14046132-14046132 on Build GRCh38
rs773431751 Chr.19: 14046264-14046264 on Build GRCh38
rs760834177 Chr.19: 14046266-14046266 on Build GRCh38

View Full Product Details