Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94488757-94488982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5875135 c.5323C>T p.R1775C Chr.8: 94488822-94488822 on Build GRCh38
1102579 c.5361C>T p.S1787S Chr.8: 94488784-94488784 on Build GRCh38
5875136 c.5308C>T p.R1770C Chr.8: 94488837-94488837 on Build GRCh38
3902144 c.5333T>C p.L1778P Chr.8: 94488812-94488812 on Build GRCh38
1102580 c.5302C>T p.R1768W Chr.8: 94488843-94488843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761062223 Chr.8: 94488873-94488873 on Build GRCh38
rs750648377 Chr.8: 94488906-94488906 on Build GRCh38
rs753108048 Chr.8: 94488867-94488867 on Build GRCh38
rs776496785 Chr.8: 94488843-94488843 on Build GRCh38
rs772649636 Chr.8: 94488941-94488941 on Build GRCh38
rs373181498 Chr.8: 94488897-94488897 on Build GRCh38
rs754329525 Chr.8: 94488885-94488885 on Build GRCh38
rs763849823 Chr.8: 94488872-94488873 on Build GRCh38
rs774321590 Chr.8: 94488877-94488878 on Build GRCh38
rs771809174 Chr.8: 94488813-94488813 on Build GRCh38
rs779033259 Chr.8: 94488784-94488784 on Build GRCh38
rs370672669 Chr.8: 94488930-94488930 on Build GRCh38
rs769309215 Chr.8: 94488848-94488848 on Build GRCh38
rs772763078 Chr.8: 94488852-94488852 on Build GRCh38
rs768572460 Chr.8: 94488838-94488838 on Build GRCh38
rs200651420 Chr.8: 94488854-94488854 on Build GRCh38
rs984479946 Chr.8: 94488878-94488878 on Build GRCh38
rs567292431 Chr.8: 94488837-94488837 on Build GRCh38
rs779789643 Chr.8: 94488814-94488814 on Build GRCh38
rs757294649 Chr.8: 94488783-94488783 on Build GRCh38
rs779025781 Chr.8: 94488903-94488903 on Build GRCh38
rs540915266 Chr.8: 94488927-94488927 on Build GRCh38
rs972376899 Chr.8: 94488821-94488821 on Build GRCh38
rs141739660 Chr.8: 94488921-94488921 on Build GRCh38
rs918282782 Chr.8: 94488936-94488936 on Build GRCh38
rs746051731 Chr.8: 94488791-94488791 on Build GRCh38
rs948377129 Chr.8: 94488904-94488904 on Build GRCh38
rs185586879 Chr.8: 94488788-94488788 on Build GRCh38
rs764701885 Chr.8: 94488883-94488883 on Build GRCh38
rs1045809695 Chr.8: 94488912-94488912 on Build GRCh38
rs76622312 Chr.8: 94488920-94488920 on Build GRCh38
rs966693554 Chr.8: 94488782-94488782 on Build GRCh38
rs747011170 Chr.8: 94488836-94488836 on Build GRCh38
rs768035032 Chr.8: 94488855-94488855 on Build GRCh38
rs533701284 Chr.8: 94488845-94488845 on Build GRCh38

View Full Product Details