Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83432141-83432406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
UniGene
Hs.480101
Gene Chromosome Location
Chr.4: 83407343-83455883 on Build GRCh38
Transcripts
NM_001297755.1 NM_001297756.1 NM_001297757.1 NM_133636.3 XM_005262711.1 XM_005262713.2 XM_006714076.2 XM_017007679.1 XM_017007680.1 XM_017007681.1 XM_017007682.1 XM_017007683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3393016 c.2126G>T p.G709V Chr.4: 83432190-83432190 on Build GRCh38
1209514 c.2128C>T p.R710C Chr.4: 83432188-83432188 on Build GRCh38
3393017 c.2125G>T p.G709C Chr.4: 83432191-83432191 on Build GRCh38
4913605 c.2065C>A p.P689T Chr.4: 83432251-83432251 on Build GRCh38
3606609 c.2065C>T p.P689S Chr.4: 83432251-83432251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761025883 Chr.4: 83432222-83432222 on Build GRCh38
rs751896103 Chr.4: 83432262-83432262 on Build GRCh38
rs774051004 Chr.4: 83432216-83432216 on Build GRCh38
rs965766536 Chr.4: 83432167-83432167 on Build GRCh38
rs756035701 Chr.4: 83432291-83432291 on Build GRCh38
rs779909482 Chr.4: 83432297-83432297 on Build GRCh38
rs1053862398 Chr.4: 83432335-83432335 on Build GRCh38
rs559204743 Chr.4: 83432239-83432239 on Build GRCh38
rs758456480 Chr.4: 83432254-83432254 on Build GRCh38
rs376507691 Chr.4: 83432270-83432270 on Build GRCh38
rs773859353 Chr.4: 83432310-83432310 on Build GRCh38
rs775889415 Chr.4: 83432266-83432267 on Build GRCh38
rs757554150 Chr.4: 83432269-83432269 on Build GRCh38
rs766664724 Chr.4: 83432224-83432224 on Build GRCh38
rs748820053 Chr.4: 83432301-83432301 on Build GRCh38
rs765695614 Chr.4: 83432251-83432251 on Build GRCh38
rs769628681 Chr.4: 83432182-83432182 on Build GRCh38
rs760039192 Chr.4: 83432241-83432241 on Build GRCh38
rs148390983 Chr.4: 83432201-83432201 on Build GRCh38
rs966500024 Chr.4: 83432316-83432316 on Build GRCh38
rs745749696 Chr.4: 83432180-83432180 on Build GRCh38
rs180763973 Chr.4: 83432187-83432187 on Build GRCh38
rs768336210 Chr.4: 83432210-83432210 on Build GRCh38
rs914938313 Chr.4: 83432366-83432366 on Build GRCh38
rs778126650 Chr.4: 83432256-83432256 on Build GRCh38
rs752790361 Chr.4: 83432253-83432253 on Build GRCh38
rs7696062 Chr.4: 83432367-83432367 on Build GRCh38
rs745667801 Chr.4: 83432286-83432286 on Build GRCh38
rs910480678 Chr.4: 83432275-83432275 on Build GRCh38

View Full Product Details