Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190360072-190360345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
UniGene
Hs.32309
Gene Chromosome Location
Chr.2: 190343470-190371665 on Build GRCh38
Transcripts
NM_001128928.1 NM_002194.3 XM_005246532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895568 c.99C>G p.I33M Chr.2: 190360201-190360201 on Build GRCh38
6310443 c.153G>T p.T51T Chr.2: 190360255-190360255 on Build GRCh38
6442734 c.112G>C p.E38Q Chr.2: 190360214-190360214 on Build GRCh38
4976268 c.147C>G p.F49L Chr.2: 190360249-190360249 on Build GRCh38
1014097 c.27C>T p.L9L Chr.2: 190360129-190360129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772047092 Chr.2: 190360133-190360133 on Build GRCh38
rs748982117 Chr.2: 190360118-190360118 on Build GRCh38
rs142570950 Chr.2: 190360169-190360169 on Build GRCh38
rs758218713 Chr.2: 190360244-190360244 on Build GRCh38
rs778908677 Chr.2: 190360254-190360254 on Build GRCh38
rs776960743 Chr.2: 190360160-190360160 on Build GRCh38
rs141092597 Chr.2: 190360316-190360316 on Build GRCh38
rs137951347 Chr.2: 190360303-190360303 on Build GRCh38
rs763357337 Chr.2: 190360296-190360296 on Build GRCh38
rs776864674 Chr.2: 190360262-190360262 on Build GRCh38
rs755218922 Chr.2: 190360230-190360230 on Build GRCh38
rs778734650 Chr.2: 190360126-190360126 on Build GRCh38
rs748017066 Chr.2: 190360127-190360127 on Build GRCh38
rs773124306 Chr.2: 190360137-190360137 on Build GRCh38
rs201135097 Chr.2: 190360258-190360258 on Build GRCh38
rs746020399 Chr.2: 190360267-190360267 on Build GRCh38
rs967894958 Chr.2: 190360232-190360232 on Build GRCh38
rs368477892 Chr.2: 190360284-190360284 on Build GRCh38
rs770963149 Chr.2: 190360158-190360158 on Build GRCh38
rs200393365 Chr.2: 190360154-190360154 on Build GRCh38
rs201566580 Chr.2: 190360195-190360195 on Build GRCh38
rs779737379 Chr.2: 190360096-190360096 on Build GRCh38
rs377606331 Chr.2: 190360312-190360312 on Build GRCh38
rs142894446 Chr.2: 190360165-190360165 on Build GRCh38
rs754582770 Chr.2: 190360314-190360314 on Build GRCh38
rs765641528 Chr.2: 190360164-190360164 on Build GRCh38
rs765875559 Chr.2: 190360181-190360181 on Build GRCh38
rs775949492 Chr.2: 190360295-190360295 on Build GRCh38
rs374089923 Chr.2: 190360202-190360202 on Build GRCh38
rs4656 Chr.2: 190360255-190360255 on Build GRCh38
rs762281676 Chr.2: 190360300-190360300 on Build GRCh38
rs917024537 Chr.2: 190360099-190360099 on Build GRCh38
rs770173204 Chr.2: 190360272-190360272 on Build GRCh38
rs112424913 Chr.2: 190360162-190360162 on Build GRCh38
rs756481569 Chr.2: 190360100-190360100 on Build GRCh38
rs2067404 Chr.2: 190360201-190360201 on Build GRCh38
rs201430776 Chr.2: 190360119-190360119 on Build GRCh38
rs369700059 Chr.2: 190360173-190360173 on Build GRCh38
rs759896532 Chr.2: 190360161-190360161 on Build GRCh38
rs755622608 Chr.2: 190360176-190360176 on Build GRCh38
rs764410591 Chr.2: 190360297-190360297 on Build GRCh38
rs747965866 Chr.2: 190360223-190360223 on Build GRCh38
rs774921776 Chr.2: 190360298-190360298 on Build GRCh38
rs753472073 Chr.2: 190360193-190360193 on Build GRCh38

View Full Product Details