Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52801418-52801685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403591 c.763G>A p.E255K Chr.5: 52801445-52801445 on Build GRCh38
4632383 c.914C>T p.A305V Chr.5: 52801596-52801596 on Build GRCh38
5770355 c.881A>G p.Y294C Chr.5: 52801563-52801563 on Build GRCh38
5130029 c.734_736delCCT p.S247delS Chr.5: 52801416-52801418 on Build GRCh38
3855481 c.801A>G p.S267S Chr.5: 52801483-52801483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041726150 Chr.5: 52801591-52801591 on Build GRCh38
rs746603199 Chr.5: 52801448-52801448 on Build GRCh38
rs755566597 Chr.5: 52801564-52801564 on Build GRCh38
rs148946513 Chr.5: 52801486-52801486 on Build GRCh38
rs373976817 Chr.5: 52801629-52801629 on Build GRCh38
rs759584138 Chr.5: 52801655-52801655 on Build GRCh38
rs563541986 Chr.5: 52801590-52801590 on Build GRCh38
rs754332528 Chr.5: 52801525-52801525 on Build GRCh38
rs758733829 Chr.5: 52801549-52801549 on Build GRCh38
rs368990050 Chr.5: 52801623-52801623 on Build GRCh38
rs773635131 Chr.5: 52801509-52801509 on Build GRCh38
rs780875619 Chr.5: 52801453-52801453 on Build GRCh38
rs927508067 Chr.5: 52801449-52801449 on Build GRCh38
rs765470395 Chr.5: 52801534-52801534 on Build GRCh38
rs749226074 Chr.5: 52801473-52801473 on Build GRCh38
rs774215540 Chr.5: 52801491-52801491 on Build GRCh38
rs749128583 Chr.5: 52801592-52801592 on Build GRCh38
rs775396002 Chr.5: 52801464-52801464 on Build GRCh38
rs778751436 Chr.5: 52801597-52801597 on Build GRCh38
rs374974514 Chr.5: 52801513-52801513 on Build GRCh38
rs755238457 Chr.5: 52801440-52801440 on Build GRCh38
rs763023657 Chr.5: 52801637-52801637 on Build GRCh38
rs756957091 Chr.5: 52801450-52801450 on Build GRCh38
rs900372558 Chr.5: 52801661-52801661 on Build GRCh38
rs745354927 Chr.5: 52801457-52801457 on Build GRCh38
rs990374199 Chr.5: 52801451-52801451 on Build GRCh38
rs765748225 Chr.5: 52801665-52801665 on Build GRCh38
rs552609052 Chr.5: 52801605-52801605 on Build GRCh38
rs767748803 Chr.5: 52801501-52801501 on Build GRCh38
rs753082949 Chr.5: 52801542-52801542 on Build GRCh38
rs975659831 Chr.5: 52801518-52801518 on Build GRCh38
rs760980313 Chr.5: 52801640-52801640 on Build GRCh38
rs776874829 Chr.5: 52801653-52801653 on Build GRCh38
rs765143574 Chr.5: 52801463-52801463 on Build GRCh38
rs755541352 Chr.5: 52801527-52801527 on Build GRCh38
rs775401633 Chr.5: 52801563-52801563 on Build GRCh38
rs1041914248 Chr.5: 52801646-52801646 on Build GRCh38
rs771096962 Chr.5: 52801644-52801644 on Build GRCh38
rs200982774 Chr.5: 52801514-52801514 on Build GRCh38
rs754185927 Chr.5: 52801583-52801583 on Build GRCh38
rs762546177 Chr.5: 52801628-52801629 on Build GRCh38
rs773508583 Chr.5: 52801631-52801631 on Build GRCh38
rs779341584 Chr.5: 52801441-52801441 on Build GRCh38
rs140897877 Chr.5: 52801594-52801594 on Build GRCh38
rs769363762 Chr.5: 52801461-52801461 on Build GRCh38
rs761558163 Chr.5: 52801498-52801498 on Build GRCh38
rs142889955 Chr.5: 52801618-52801618 on Build GRCh38
rs879227586 Chr.5: 52801649-52801649 on Build GRCh38

View Full Product Details