Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33166585-33166858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A2
Entrez Gene ID
1302
Gene Name
collagen type XI alpha 2
Gene Aliases
DFNA13, DFNB53, FBCG2, HKE5, PARP, STL3
UniGene
Hs.390171
Gene Chromosome Location
Chr.6: 33162692-33193009 on Build GRCh38
Transcripts
NM_080679.2 NM_080680.2 NM_080681.2 XM_011514299.2 XM_011514300.2 XM_011514302.2 XM_017010250.1 XM_017010251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921508 c.4249G>A p.G1417R Chr.6: 33166809-33166809 on Build GRCh38
399541 c.4237C>A p.P1413T Chr.6: 33166821-33166821 on Build GRCh38
3625605 c.4312G>A p.G1438S Chr.6: 33166746-33166746 on Build GRCh38
137453 c.4238C>T p.P1413L Chr.6: 33166820-33166820 on Build GRCh38
4006451 c.4266G>A p.P1422P Chr.6: 33166792-33166792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765708674 Chr.6: 33166803-33166803 on Build GRCh38
rs918752772 Chr.6: 33166823-33166823 on Build GRCh38
rs371596685 Chr.6: 33166619-33166619 on Build GRCh38
rs773474660 Chr.6: 33166788-33166788 on Build GRCh38
rs754334343 Chr.6: 33166798-33166798 on Build GRCh38
rs200408948 Chr.6: 33166612-33166612 on Build GRCh38
rs573864576 Chr.6: 33166634-33166634 on Build GRCh38
rs757898834 Chr.6: 33166824-33166824 on Build GRCh38
rs774521995 Chr.6: 33166740-33166740 on Build GRCh38
rs2855453 Chr.6: 33166615-33166615 on Build GRCh38
rs985479138 Chr.6: 33166770-33166770 on Build GRCh38
rs1003092656 Chr.6: 33166645-33166645 on Build GRCh38
rs375023453 Chr.6: 33166699-33166699 on Build GRCh38
rs371538774 Chr.6: 33166698-33166698 on Build GRCh38
rs761955347 Chr.6: 33166766-33166766 on Build GRCh38
rs1048004384 Chr.6: 33166633-33166633 on Build GRCh38
rs768885036 Chr.6: 33166739-33166739 on Build GRCh38
rs752370087 Chr.6: 33166675-33166675 on Build GRCh38
rs752243096 Chr.6: 33166806-33166806 on Build GRCh38
rs758093145 Chr.6: 33166685-33166685 on Build GRCh38
rs555936333 Chr.6: 33166793-33166793 on Build GRCh38
rs564087840 Chr.6: 33166831-33166831 on Build GRCh38
rs757461695 Chr.6: 33166629-33166629 on Build GRCh38
rs745747882 Chr.6: 33166733-33166733 on Build GRCh38
rs760960909 Chr.6: 33166792-33166792 on Build GRCh38
rs751161439 Chr.6: 33166837-33166837 on Build GRCh38
rs865979749 Chr.6: 33166781-33166781 on Build GRCh38
rs781001180 Chr.6: 33166721-33166721 on Build GRCh38
rs759989401 Chr.6: 33166802-33166802 on Build GRCh38
rs121912946 Chr.6: 33166736-33166736 on Build GRCh38
rs955878484 Chr.6: 33166689-33166689 on Build GRCh38
rs982696708 Chr.6: 33166607-33166607 on Build GRCh38
rs369126897 Chr.6: 33166767-33166767 on Build GRCh38
rs1031493071 Chr.6: 33166656-33166656 on Build GRCh38
rs747979390 Chr.6: 33166649-33166649 on Build GRCh38
rs777372477 Chr.6: 33166694-33166694 on Build GRCh38
rs554694195 Chr.6: 33166628-33166628 on Build GRCh38

View Full Product Details