Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74592647-74592894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
-
UniGene
Hs.350775
Gene Chromosome Location
Chr.17: 74584679-74594209 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5668060 c.725G>T p.G242V Chr.17: 74592771-74592771 on Build GRCh38
3717586 c.695T>C p.L232P Chr.17: 74592741-74592741 on Build GRCh38
6481693 c.668C>G p.S223C Chr.17: 74592714-74592714 on Build GRCh38
3890351 c.672C>T p.S224S Chr.17: 74592718-74592718 on Build GRCh38
5678914 c.604G>T p.A202S Chr.17: 74592650-74592650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140166356 Chr.17: 74592761-74592761 on Build GRCh38
rs751224082 Chr.17: 74592731-74592731 on Build GRCh38
rs867736187 Chr.17: 74592765-74592765 on Build GRCh38
rs773560087 Chr.17: 74592722-74592722 on Build GRCh38
rs768849227 Chr.17: 74592873-74592873 on Build GRCh38
rs146127120 Chr.17: 74592760-74592760 on Build GRCh38
rs1012772501 Chr.17: 74592865-74592865 on Build GRCh38
rs373006665 Chr.17: 74592826-74592826 on Build GRCh38
rs770429182 Chr.17: 74592808-74592808 on Build GRCh38
rs988872728 Chr.17: 74592767-74592767 on Build GRCh38
rs375772119 Chr.17: 74592788-74592788 on Build GRCh38
rs778964245 Chr.17: 74592864-74592864 on Build GRCh38
rs144355502 Chr.17: 74592754-74592754 on Build GRCh38
rs150862645 Chr.17: 74592729-74592729 on Build GRCh38
rs751394899 Chr.17: 74592683-74592683 on Build GRCh38
rs545652 Chr.17: 74592667-74592667 on Build GRCh38
rs756990289 Chr.17: 74592743-74592743 on Build GRCh38
rs755837698 Chr.17: 74592744-74592744 on Build GRCh38
rs577395405 Chr.17: 74592787-74592787 on Build GRCh38
rs767146842 Chr.17: 74592741-74592741 on Build GRCh38
rs777798292 Chr.17: 74592746-74592746 on Build GRCh38
rs191930489 Chr.17: 74592689-74592689 on Build GRCh38
rs767484916 Chr.17: 74592795-74592795 on Build GRCh38
rs890956654 Chr.17: 74592866-74592866 on Build GRCh38
rs369351414 Chr.17: 74592679-74592679 on Build GRCh38
rs915459221 Chr.17: 74592708-74592708 on Build GRCh38
rs753670280 Chr.17: 74592748-74592748 on Build GRCh38
rs747170192 Chr.17: 74592698-74592698 on Build GRCh38
rs1029823807 Chr.17: 74592691-74592692 on Build GRCh38
rs557704605 Chr.17: 74592730-74592730 on Build GRCh38
rs373075685 Chr.17: 74592755-74592755 on Build GRCh38
rs369536809 Chr.17: 74592801-74592801 on Build GRCh38
rs760507571 Chr.17: 74592714-74592714 on Build GRCh38
rs773787386 Chr.17: 74592796-74592796 on Build GRCh38
rs759377407 Chr.17: 74592788-74592792 on Build GRCh38
rs772007436 Chr.17: 74592676-74592676 on Build GRCh38
rs747048899 Chr.17: 74592766-74592766 on Build GRCh38
rs754879895 Chr.17: 74592830-74592830 on Build GRCh38
rs753753271 Chr.17: 74592819-74592819 on Build GRCh38
rs139358664 Chr.17: 74592749-74592749 on Build GRCh38
rs759255401 Chr.17: 74592739-74592739 on Build GRCh38
rs762071099 Chr.17: 74592706-74592706 on Build GRCh38
rs769148474 Chr.17: 74592701-74592701 on Build GRCh38
rs752541699 Chr.17: 74592742-74592742 on Build GRCh38
rs777202016 Chr.17: 74592703-74592703 on Build GRCh38

View Full Product Details