Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46813735-46813992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4B1
Entrez Gene ID
1580
Gene Name
cytochrome P450 family 4 subfamily B member 1
Gene Aliases
CYPIVB1, P-450HP
UniGene
Hs.436317
Gene Chromosome Location
Chr.1: 46798995-46819349 on Build GRCh38
Transcripts
NM_000779.3 NM_001099772.1 NM_001319161.1 NM_001319162.1 NM_001319163.1 NR_135003.1 XM_011540832.1 XM_011540833.1 XM_017000466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490358 c.621G>A p.R207R Chr.1: 46813912-46813912 on Build GRCh38
2191446 c.633C>A p.Y211* Chr.1: 46813924-46813924 on Build GRCh38
6849207 c.644T>C p.V215A Chr.1: 46813935-46813935 on Build GRCh38
4008611 c.673C>T p.R225C Chr.1: 46813964-46813964 on Build GRCh38
3490359 c.676C>T p.L226F Chr.1: 46813967-46813967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55826230 Chr.1: 46813852-46813852 on Build GRCh38
rs751225806 Chr.1: 46813888-46813888 on Build GRCh38
rs201496388 Chr.1: 46813901-46813901 on Build GRCh38
rs763795819 Chr.1: 46813876-46813876 on Build GRCh38
rs45584834 Chr.1: 46813824-46813824 on Build GRCh38
rs188313819 Chr.1: 46813909-46813909 on Build GRCh38
rs757516153 Chr.1: 46813845-46813845 on Build GRCh38
rs201769002 Chr.1: 46813894-46813896 on Build GRCh38
rs397822024 Chr.1: 46813893-46813895 on Build GRCh38
rs943795661 Chr.1: 46813878-46813878 on Build GRCh38
rs534832348 Chr.1: 46813822-46813822 on Build GRCh38
rs1027204040 Chr.1: 46813813-46813813 on Build GRCh38
rs779008485 Chr.1: 46813930-46813930 on Build GRCh38
rs376687142 Chr.1: 46813956-46813956 on Build GRCh38
rs17102575 Chr.1: 46813760-46813760 on Build GRCh38
rs370030027 Chr.1: 46813903-46813903 on Build GRCh38
rs55774598 Chr.1: 46813821-46813821 on Build GRCh38
rs144659997 Chr.1: 46813964-46813964 on Build GRCh38
rs139750942 Chr.1: 46813940-46813940 on Build GRCh38
rs71541955 Chr.1: 46813847-46813847 on Build GRCh38
rs575412611 Chr.1: 46813897-46813897 on Build GRCh38
rs771290125 Chr.1: 46813945-46813945 on Build GRCh38
rs45527338 Chr.1: 46813789-46813789 on Build GRCh38
rs772342186 Chr.1: 46813939-46813939 on Build GRCh38
rs981115756 Chr.1: 46813783-46813783 on Build GRCh38
rs200579490 Chr.1: 46813890-46813890 on Build GRCh38
rs747410215 Chr.1: 46813942-46813942 on Build GRCh38
rs372304855 Chr.1: 46813862-46813862 on Build GRCh38
rs750366022 Chr.1: 46813965-46813965 on Build GRCh38
rs368797537 Chr.1: 46813961-46813961 on Build GRCh38
rs537269144 Chr.1: 46813785-46813785 on Build GRCh38
rs201819214 Chr.1: 46813886-46813888 on Build GRCh38
rs980714398 Chr.1: 46813808-46813808 on Build GRCh38
rs75581734 Chr.1: 46813839-46813839 on Build GRCh38
rs200120700 Chr.1: 46813927-46813927 on Build GRCh38
rs748352207 Chr.1: 46813935-46813935 on Build GRCh38
rs908742299 Chr.1: 46813833-46813833 on Build GRCh38
rs56392063 Chr.1: 46813804-46813804 on Build GRCh38
rs4646489 Chr.1: 46813887-46813889 on Build GRCh38
rs61011955 Chr.1: 46813920-46813920 on Build GRCh38
rs931653870 Chr.1: 46813899-46813899 on Build GRCh38
rs760151276 Chr.1: 46813964-46813965 on Build GRCh38

View Full Product Details