Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2232607-2232868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E4F1
Entrez Gene ID
1877
Gene Name
E4F transcription factor 1
Gene Aliases
E4F
UniGene
Hs.513268
Gene Chromosome Location
Chr.16: 2223488-2235742 on Build GRCh38
Transcripts
NM_001288776.1 NM_001288778.1 NM_004424.4 XM_005255155.1 XM_005255156.1 XM_006720858.1 XM_011522402.1 XM_011522403.1 XM_017023010.1 XM_017023011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015346 c.838A>G p.S280G Chr.16: 2232863-2232863 on Build GRCh38
4059319 c.737G>A p.R246H Chr.16: 2232762-2232762 on Build GRCh38
74475 c.757T>G p.C253G Chr.16: 2232782-2232782 on Build GRCh38
6021397 c.831C>G p.I277M Chr.16: 2232856-2232856 on Build GRCh38
4059320 c.780G>A p.S260S Chr.16: 2232805-2232805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139438270 Chr.16: 2232805-2232805 on Build GRCh38
rs757045675 Chr.16: 2232710-2232710 on Build GRCh38
rs771304579 Chr.16: 2232772-2232772 on Build GRCh38
rs899438778 Chr.16: 2232636-2232636 on Build GRCh38
rs995088060 Chr.16: 2232638-2232638 on Build GRCh38
rs755060372 Chr.16: 2232748-2232748 on Build GRCh38
rs947191420 Chr.16: 2232658-2232680 on Build GRCh38
rs994404438 Chr.16: 2232641-2232641 on Build GRCh38
rs893264993 Chr.16: 2232673-2232673 on Build GRCh38
rs748369000 Chr.16: 2232757-2232757 on Build GRCh38
rs372896150 Chr.16: 2232752-2232752 on Build GRCh38
rs369411860 Chr.16: 2232751-2232751 on Build GRCh38
rs370499012 Chr.16: 2232818-2232818 on Build GRCh38
rs1030691504 Chr.16: 2232703-2232703 on Build GRCh38
rs199803798 Chr.16: 2232786-2232786 on Build GRCh38
rs1001733387 Chr.16: 2232671-2232671 on Build GRCh38
rs924472431 Chr.16: 2232839-2232839 on Build GRCh38
rs765020785 Chr.16: 2232808-2232808 on Build GRCh38
rs749705386 Chr.16: 2232769-2232769 on Build GRCh38
rs758235332 Chr.16: 2232724-2232724 on Build GRCh38
rs934521424 Chr.16: 2232846-2232846 on Build GRCh38
rs941752803 Chr.16: 2232787-2232787 on Build GRCh38
rs1034426566 Chr.16: 2232672-2232672 on Build GRCh38
rs113710566 Chr.16: 2232698-2232698 on Build GRCh38
rs1026922279 Chr.16: 2232646-2232646 on Build GRCh38
rs967800325 Chr.16: 2232683-2232683 on Build GRCh38
rs753559375 Chr.16: 2232709-2232709 on Build GRCh38
rs528812715 Chr.16: 2232750-2232750 on Build GRCh38
rs1011676096 Chr.16: 2232674-2232674 on Build GRCh38
rs773429419 Chr.16: 2232764-2232764 on Build GRCh38
rs886607709 Chr.16: 2232647-2232647 on Build GRCh38
rs750279613 Chr.16: 2232719-2232719 on Build GRCh38
rs1020255049 Chr.16: 2232682-2232682 on Build GRCh38
rs140166330 Chr.16: 2232775-2232775 on Build GRCh38
rs375194455 Chr.16: 2232833-2232833 on Build GRCh38
rs770063253 Chr.16: 2232762-2232762 on Build GRCh38
rs780070482 Chr.16: 2232727-2232727 on Build GRCh38
rs566348970 Chr.16: 2232730-2232730 on Build GRCh38
rs750143067 Chr.16: 2232816-2232816 on Build GRCh38
rs763701964 Chr.16: 2232797-2232797 on Build GRCh38
rs552394617 Chr.16: 2232694-2232694 on Build GRCh38
rs776338481 Chr.16: 2232804-2232804 on Build GRCh38

View Full Product Details