Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45137751-45138018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001135705.2 NM_001135706.2 NM_001135707.2 NM_001321352.1 NM_001321353.1 NM_024722.3 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025094.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3191991 c.589C>T p.R197W Chr.17: 45137928-45137928 on Build GRCh38
3191990 c.627C>T p.S209S Chr.17: 45137928-45137928 on Build GRCh38
6571464 c.611+3G>A p.? Chr.17: 45137833-45137833 on Build GRCh38
5544406 c.549C>T p.S183S Chr.17: 45137806-45137806 on Build GRCh38
4381871 c.552C>A p.H184Q Chr.17: 45137771-45137771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772977455 Chr.17: 45137949-45137949 on Build GRCh38
rs756264985 Chr.17: 45137903-45137903 on Build GRCh38
rs371250075 Chr.17: 45137877-45137877 on Build GRCh38
rs530738927 Chr.17: 45137864-45137864 on Build GRCh38
rs768396117 Chr.17: 45137924-45137924 on Build GRCh38
rs368818663 Chr.17: 45137872-45137872 on Build GRCh38
rs1008484950 Chr.17: 45137889-45137889 on Build GRCh38
rs766958779 Chr.17: 45137863-45137863 on Build GRCh38
rs772473601 Chr.17: 45137979-45137979 on Build GRCh38
rs561115754 Chr.17: 45137852-45137852 on Build GRCh38
rs990895706 Chr.17: 45137931-45137931 on Build GRCh38
rs867143035 Chr.17: 45137776-45137776 on Build GRCh38
rs546766187 Chr.17: 45137928-45137928 on Build GRCh38
rs761479549 Chr.17: 45137855-45137855 on Build GRCh38
rs541746362 Chr.17: 45137844-45137844 on Build GRCh38
rs765650791 Chr.17: 45137871-45137871 on Build GRCh38
rs541161009 Chr.17: 45137801-45137801 on Build GRCh38
rs771708890 Chr.17: 45137940-45137940 on Build GRCh38
rs199961151 Chr.17: 45137926-45137926 on Build GRCh38
rs200905184 Chr.17: 45137970-45137970 on Build GRCh38
rs3830314 Chr.17: 45137900-45137904 on Build GRCh38
rs753002354 Chr.17: 45137797-45137797 on Build GRCh38
rs779272057 Chr.17: 45137789-45137789 on Build GRCh38
rs757555935 Chr.17: 45137883-45137883 on Build GRCh38
rs574537572 Chr.17: 45137842-45137842 on Build GRCh38
rs764681492 Chr.17: 45137848-45137849 on Build GRCh38
rs760081117 Chr.17: 45137950-45137950 on Build GRCh38
rs202115808 Chr.17: 45137853-45137853 on Build GRCh38
rs564309161 Chr.17: 45137917-45137917 on Build GRCh38
rs528174632 Chr.17: 45137918-45137918 on Build GRCh38
rs773519074 Chr.17: 45137987-45137987 on Build GRCh38
rs1015471048 Chr.17: 45137891-45137891 on Build GRCh38
rs781627367 Chr.17: 45137887-45137887 on Build GRCh38
rs545826995 Chr.17: 45137772-45137772 on Build GRCh38
rs775553830 Chr.17: 45137850-45137850 on Build GRCh38
rs918006056 Chr.17: 45137963-45137963 on Build GRCh38
rs375328317 Chr.17: 45137955-45137955 on Build GRCh38
rs775810190 Chr.17: 45137956-45137956 on Build GRCh38
rs892874816 Chr.17: 45137847-45137847 on Build GRCh38
rs996633286 Chr.17: 45137851-45137851 on Build GRCh38
rs778120528 Chr.17: 45137807-45137807 on Build GRCh38
rs747247734 Chr.17: 45137948-45137948 on Build GRCh38
rs561659128 Chr.17: 45137929-45137929 on Build GRCh38
rs751773949 Chr.17: 45137878-45137878 on Build GRCh38
rs771086570 Chr.17: 45137838-45137838 on Build GRCh38
rs1054086507 Chr.17: 45137803-45137803 on Build GRCh38
rs755457256 Chr.17: 45137788-45137788 on Build GRCh38
rs764485010 Chr.17: 45137972-45137972 on Build GRCh38
rs745919135 Chr.17: 45137893-45137893 on Build GRCh38
rs767610226 Chr.17: 45137994-45137994 on Build GRCh38
rs886316132 Chr.17: 45137983-45137983 on Build GRCh38
rs374955857 Chr.17: 45137909-45137909 on Build GRCh38
rs552925104 Chr.17: 45137825-45137825 on Build GRCh38
rs751965924 Chr.17: 45137978-45137978 on Build GRCh38
rs769610747 Chr.17: 45137849-45137849 on Build GRCh38
rs375037904 Chr.17: 45137865-45137865 on Build GRCh38

View Full Product Details