Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114439079-114439352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
UniGene
Hs.438236
Gene Chromosome Location
Chr.10: 114431110-114821643 on Build GRCh38
Transcripts
NM_001003407.1 NM_001003408.1 NM_001322882.1 NM_001322883.1 NM_001322884.1 NM_001322885.1 NM_001322886.1 NM_001322887.1 NM_001322888.1 NM_001322889.1 NM_001322890.1 NM_001322891.1 NM_001322892.1 NM_001322893.1 NM_001322894.1 NM_001322895.1 NM_001322896.1 NM_001322897.1 NM_001322898.1 NM_001322899.1 NM_001322900.1 NM_002313.5 NM_006720.3 XM_005269824.2 XM_005269826.1 XM_005269827.4 XM_006717837.1 XM_006717846.3 XM_006717847.1 XM_011539801.1 XM_011539802.1 XM_011539803.1 XM_011539805.1 XM_017016242.1 XM_017016243.1 XM_017016244.1 XM_017016245.1 XM_017016246.1 XM_017016247.1 XM_017016248.1 XM_017016249.1 XM_017016250.1 XM_017016251.1 XM_017016252.1 XM_017016253.1 XM_017016254.1 XM_017016255.1 XM_017016256.1 XM_017016258.1 XM_017016259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011796 c.1130G>T p.R377I Chr.10: 114439219-114439219 on Build GRCh38
3709760 c.2319G>A p.V773V Chr.10: 114439203-114439203 on Build GRCh38
4011792 c.1139G>A p.R380Q Chr.10: 114439210-114439210 on Build GRCh38
3709758 c.1935G>A p.V645V Chr.10: 114439203-114439203 on Build GRCh38
3709757 c.2115G>A p.V705V Chr.10: 114439203-114439203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768916518 Chr.10: 114439213-114439213 on Build GRCh38
rs558984043 Chr.10: 114439116-114439116 on Build GRCh38
rs775062919 Chr.10: 114439243-114439243 on Build GRCh38
rs772917489 Chr.10: 114439219-114439219 on Build GRCh38
rs377280713 Chr.10: 114439288-114439288 on Build GRCh38
rs189596395 Chr.10: 114439169-114439169 on Build GRCh38
rs776670411 Chr.10: 114439232-114439232 on Build GRCh38
rs780339538 Chr.10: 114439210-114439210 on Build GRCh38
rs139367133 Chr.10: 114439237-114439237 on Build GRCh38
rs184729101 Chr.10: 114439153-114439153 on Build GRCh38
rs779147136 Chr.10: 114439306-114439306 on Build GRCh38
rs1046561088 Chr.10: 114439203-114439203 on Build GRCh38
rs749527326 Chr.10: 114439211-114439211 on Build GRCh38
rs756597263 Chr.10: 114439293-114439293 on Build GRCh38
rs781631215 Chr.10: 114439181-114439181 on Build GRCh38
rs367814605 Chr.10: 114439146-114439146 on Build GRCh38
rs1032444315 Chr.10: 114439244-114439244 on Build GRCh38
rs746830805 Chr.10: 114439310-114439310 on Build GRCh38
rs746794140 Chr.10: 114439223-114439223 on Build GRCh38
rs545246696 Chr.10: 114439305-114439305 on Build GRCh38
rs751894855 Chr.10: 114439264-114439264 on Build GRCh38
rs780061185 Chr.10: 114439297-114439297 on Build GRCh38
rs771015001 Chr.10: 114439224-114439224 on Build GRCh38
rs369362851 Chr.10: 114439147-114439147 on Build GRCh38
rs375311883 Chr.10: 114439135-114439135 on Build GRCh38
rs1020194549 Chr.10: 114439265-114439265 on Build GRCh38
rs764435013 Chr.10: 114439167-114439167 on Build GRCh38
rs374666555 Chr.10: 114439274-114439274 on Build GRCh38
rs778253601 Chr.10: 114439288-114439292 on Build GRCh38
rs745982928 Chr.10: 114439194-114439194 on Build GRCh38
rs768018436 Chr.10: 114439284-114439284 on Build GRCh38
rs756123104 Chr.10: 114439200-114439200 on Build GRCh38
rs757811100 Chr.10: 114439170-114439170 on Build GRCh38
rs371358908 Chr.10: 114439258-114439258 on Build GRCh38
rs372490861 Chr.10: 114439161-114439161 on Build GRCh38
rs958048282 Chr.10: 114439255-114439255 on Build GRCh38
rs201622805 Chr.10: 114439136-114439136 on Build GRCh38
rs376194277 Chr.10: 114439249-114439249 on Build GRCh38
rs765012284 Chr.10: 114439137-114439137 on Build GRCh38
rs770780381 Chr.10: 114439311-114439311 on Build GRCh38

View Full Product Details