Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32357403-32357668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
UniGene
Hs.150107
Gene Chromosome Location
Chr.2: 32356907-32618899 on Build GRCh38
Transcripts
NM_016252.3 XM_005264449.4 XM_005264450.4 XM_005264451.4 XM_005264452.4 XM_005264453.4 XM_005264454.4 XM_005264455.4 XM_006712054.3 XM_006712055.3 XM_006712056.3 XM_011533005.2 XM_017004556.1 XM_017004557.1 XM_017004558.1 XM_017004559.1 XM_017004560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391678 c.301A>T p.T101S Chr.2: 32357462-32357462 on Build GRCh38
6996340 c.250G>A p.A84T Chr.2: 32357411-32357411 on Build GRCh38
7240733 c.273C>T p.I91I Chr.2: 32357434-32357434 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542493800 Chr.2: 32357607-32357607 on Build GRCh38
rs753784378 Chr.2: 32357465-32357465 on Build GRCh38
rs1031131586 Chr.2: 32357438-32357438 on Build GRCh38
rs761446695 Chr.2: 32357482-32357482 on Build GRCh38
rs892840893 Chr.2: 32357640-32357645 on Build GRCh38
rs116200779 Chr.2: 32357587-32357587 on Build GRCh38
rs1050662070 Chr.2: 32357597-32357597 on Build GRCh38
rs896157227 Chr.2: 32357582-32357582 on Build GRCh38
rs1000945638 Chr.2: 32357424-32357424 on Build GRCh38
rs1003121893 Chr.2: 32357519-32357519 on Build GRCh38
rs984723621 Chr.2: 32357558-32357558 on Build GRCh38
rs903686707 Chr.2: 32357598-32357598 on Build GRCh38
rs556087079 Chr.2: 32357625-32357625 on Build GRCh38
rs923310018 Chr.2: 32357509-32357509 on Build GRCh38
rs1038079460 Chr.2: 32357557-32357557 on Build GRCh38
rs942235405 Chr.2: 32357546-32357546 on Build GRCh38
rs1000724712 Chr.2: 32357645-32357645 on Build GRCh38
rs929643766 Chr.2: 32357564-32357564 on Build GRCh38
rs916620558 Chr.2: 32357532-32357532 on Build GRCh38
rs958901467 Chr.2: 32357521-32357521 on Build GRCh38
rs1040953408 Chr.2: 32357574-32357574 on Build GRCh38
rs761789741 Chr.2: 32357502-32357517 on Build GRCh38
rs750154381 Chr.2: 32357484-32357484 on Build GRCh38
rs1045039651 Chr.2: 32357596-32357596 on Build GRCh38
rs191125788 Chr.2: 32357573-32357573 on Build GRCh38
rs758064107 Chr.2: 32357497-32357497 on Build GRCh38
rs986402419 Chr.2: 32357536-32357536 on Build GRCh38
rs918358260 Chr.2: 32357562-32357562 on Build GRCh38
rs909350937 Chr.2: 32357541-32357541 on Build GRCh38
rs932248120 Chr.2: 32357593-32357593 on Build GRCh38
rs991404659 Chr.2: 32357528-32357528 on Build GRCh38
rs764814191 Chr.2: 32357483-32357483 on Build GRCh38
rs534828568 Chr.2: 32357543-32357543 on Build GRCh38
rs976519327 Chr.2: 32357504-32357504 on Build GRCh38
rs779506409 Chr.2: 32357520-32357520 on Build GRCh38

View Full Product Details