Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 21001784-21002053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOB
Entrez Gene ID
338
Gene Name
apolipoprotein B
Gene Aliases
FLDB, LDLCQ4, apoB-100, apoB-48
UniGene
Hs.120759
Gene Chromosome Location
Chr.2: 21001429-21044073 on Build GRCh38
Transcripts
NM_000384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015886 c.13399G>T p.A4467S Chr.2: 21002023-21002023 on Build GRCh38
1015883 c.13476C>T p.H4492H Chr.2: 21001946-21001946 on Build GRCh38
1405049 c.13527C>A p.Y4509* Chr.2: 21001895-21001895 on Build GRCh38
1015882 c.13538T>G p.F4513C Chr.2: 21001884-21001884 on Build GRCh38
7324023 c.13487G>A p.R4496K Chr.2: 21001935-21001935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776276258 Chr.2: 21001977-21001977 on Build GRCh38
rs749634224 Chr.2: 21001937-21001937 on Build GRCh38
rs201587166 Chr.2: 21002006-21002006 on Build GRCh38
rs756545438 Chr.2: 21001939-21001939 on Build GRCh38
rs146661220 Chr.2: 21002000-21002000 on Build GRCh38
rs377641527 Chr.2: 21001814-21001814 on Build GRCh38
rs779345915 Chr.2: 21002018-21002018 on Build GRCh38
rs770544459 Chr.2: 21001853-21001853 on Build GRCh38
rs200374122 Chr.2: 21001970-21001970 on Build GRCh38
rs900908225 Chr.2: 21002028-21002028 on Build GRCh38
rs1021009945 Chr.2: 21001864-21001864 on Build GRCh38
rs574659380 Chr.2: 21002013-21002013 on Build GRCh38
rs770231338 Chr.2: 21001841-21001841 on Build GRCh38
rs778140846 Chr.2: 21001833-21001833 on Build GRCh38
rs757136007 Chr.2: 21001915-21001915 on Build GRCh38
rs756787301 Chr.2: 21001901-21001902 on Build GRCh38
rs756281873 Chr.2: 21002002-21002002 on Build GRCh38
rs138421941 Chr.2: 21001973-21001973 on Build GRCh38
rs781520074 Chr.2: 21001936-21001936 on Build GRCh38
rs12713450 Chr.2: 21001971-21001971 on Build GRCh38
rs897737982 Chr.2: 21001817-21001817 on Build GRCh38
rs771143412 Chr.2: 21001946-21001946 on Build GRCh38
rs374389311 Chr.2: 21001959-21001959 on Build GRCh38
rs753852142 Chr.2: 21001892-21001892 on Build GRCh38
rs141477444 Chr.2: 21001922-21001922 on Build GRCh38
rs562574661 Chr.2: 21001940-21001942 on Build GRCh38
rs749549578 Chr.2: 21002010-21002010 on Build GRCh38
rs546806987 Chr.2: 21001867-21001867 on Build GRCh38
rs748453503 Chr.2: 21001839-21001839 on Build GRCh38
rs755251454 Chr.2: 21001934-21001934 on Build GRCh38
rs748727595 Chr.2: 21001815-21001815 on Build GRCh38
rs773822083 Chr.2: 21001842-21001842 on Build GRCh38
rs749868276 Chr.2: 21001965-21001965 on Build GRCh38
rs371437581 Chr.2: 21001856-21001856 on Build GRCh38
rs763773989 Chr.2: 21001872-21001872 on Build GRCh38
rs771286073 Chr.2: 21001968-21001968 on Build GRCh38
rs886055571 Chr.2: 21001821-21001821 on Build GRCh38
rs938535592 Chr.2: 21001818-21001818 on Build GRCh38
rs751629013 Chr.2: 21001933-21001933 on Build GRCh38
rs774487085 Chr.2: 21001966-21001966 on Build GRCh38
rs763341676 Chr.2: 21001846-21001846 on Build GRCh38
rs1801695 Chr.2: 21001981-21001981 on Build GRCh38
rs566760356 Chr.2: 21001919-21001919 on Build GRCh38
rs767091894 Chr.2: 21001858-21001858 on Build GRCh38
rs756692121 Chr.2: 21001828-21001828 on Build GRCh38
rs755057102 Chr.2: 21001982-21001982 on Build GRCh38
rs370286693 Chr.2: 21001976-21001976 on Build GRCh38
rs112667751 Chr.2: 21001905-21001905 on Build GRCh38
rs764014607 Chr.2: 21001975-21001975 on Build GRCh38
rs767676919 Chr.2: 21001995-21001995 on Build GRCh38
rs142702699 Chr.2: 21001978-21001978 on Build GRCh38
rs773841094 Chr.2: 21001855-21001855 on Build GRCh38
rs958014928 Chr.2: 21001836-21001836 on Build GRCh38
rs147416761 Chr.2: 21001974-21001974 on Build GRCh38
rs760551634 Chr.2: 21002011-21002011 on Build GRCh38

View Full Product Details