Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88807354-88807579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDT1
Entrez Gene ID
81620
Gene Name
chromatin licensing and DNA replication factor 1
Gene Aliases
DUP, RIS2
UniGene
Hs.122908
Gene Chromosome Location
Chr.16: 88803778-88809258 on Build GRCh38
Transcripts
NM_030928.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851691 c.1454G>A p.S485N Chr.16: 88807459-88807459 on Build GRCh38
3969744 c.1349G>A p.R450H Chr.16: 88807354-88807354 on Build GRCh38
5620463 c.1385G>A p.R462Q Chr.16: 88807390-88807390 on Build GRCh38
5701102 c.1362_1363insG p.P455fs*2 Chr.16: 88807367-88807368 on Build GRCh38
4579363 c.1383delG p.R462fs*29 Chr.16: 88807388-88807388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781732972 Chr.16: 88807432-88807433 on Build GRCh38
rs751213474 Chr.16: 88807381-88807381 on Build GRCh38
rs374693189 Chr.16: 88807500-88807500 on Build GRCh38
rs760038187 Chr.16: 88807435-88807435 on Build GRCh38
rs760255561 Chr.16: 88807495-88807495 on Build GRCh38
rs908267383 Chr.16: 88807549-88807549 on Build GRCh38
rs769923406 Chr.16: 88807486-88807507 on Build GRCh38
rs757547495 Chr.16: 88807506-88807506 on Build GRCh38
rs377536198 Chr.16: 88807508-88807508 on Build GRCh38
rs760114445 Chr.16: 88807450-88807450 on Build GRCh38
rs151292317 Chr.16: 88807406-88807406 on Build GRCh38
rs769002955 Chr.16: 88807398-88807399 on Build GRCh38
rs747989537 Chr.16: 88807527-88807527 on Build GRCh38
rs753096632 Chr.16: 88807439-88807439 on Build GRCh38
rs755500544 Chr.16: 88807389-88807389 on Build GRCh38
rs933332385 Chr.16: 88807397-88807397 on Build GRCh38
rs749966576 Chr.16: 88807429-88807429 on Build GRCh38
rs771038378 Chr.16: 88807405-88807405 on Build GRCh38
rs201281960 Chr.16: 88807496-88807496 on Build GRCh38
rs550557201 Chr.16: 88807470-88807470 on Build GRCh38
rs765805425 Chr.16: 88807438-88807438 on Build GRCh38
rs781210319 Chr.16: 88807454-88807454 on Build GRCh38
rs749300368 Chr.16: 88807519-88807519 on Build GRCh38
rs747209207 Chr.16: 88807400-88807400 on Build GRCh38
rs202071237 Chr.16: 88807420-88807420 on Build GRCh38
rs756263215 Chr.16: 88807514-88807514 on Build GRCh38
rs567319280 Chr.16: 88807524-88807524 on Build GRCh38
rs960242606 Chr.16: 88807504-88807504 on Build GRCh38
rs757456654 Chr.16: 88807449-88807449 on Build GRCh38
rs147704737 Chr.16: 88807474-88807474 on Build GRCh38
rs541685901 Chr.16: 88807553-88807554 on Build GRCh38
rs763839264 Chr.16: 88807380-88807380 on Build GRCh38
rs751844135 Chr.16: 88807448-88807448 on Build GRCh38
rs771857191 Chr.16: 88807486-88807486 on Build GRCh38
rs370178302 Chr.16: 88807522-88807522 on Build GRCh38
rs749210361 Chr.16: 88807475-88807475 on Build GRCh38
rs145394645 Chr.16: 88807378-88807378 on Build GRCh38
rs377758402 Chr.16: 88807487-88807487 on Build GRCh38
rs146199695 Chr.16: 88807477-88807477 on Build GRCh38
rs767196625 Chr.16: 88807422-88807422 on Build GRCh38
rs948767391 Chr.16: 88807403-88807403 on Build GRCh38
rs141750196 Chr.16: 88807410-88807410 on Build GRCh38
rs778080229 Chr.16: 88807447-88807447 on Build GRCh38
rs748552177 Chr.16: 88807392-88807392 on Build GRCh38
rs3218729 Chr.16: 88807372-88807372 on Build GRCh38
rs761646387 Chr.16: 88807483-88807483 on Build GRCh38
rs758722232 Chr.16: 88807395-88807395 on Build GRCh38
rs115396767 Chr.16: 88807534-88807534 on Build GRCh38
rs387906917 Chr.16: 88807390-88807390 on Build GRCh38
rs779997523 Chr.16: 88807515-88807515 on Build GRCh38
rs748117647 Chr.16: 88807470-88807471 on Build GRCh38
rs765007374 Chr.16: 88807374-88807374 on Build GRCh38
rs141663679 Chr.16: 88807416-88807416 on Build GRCh38
rs3218730 Chr.16: 88807382-88807382 on Build GRCh38
rs777993968 Chr.16: 88807399-88807399 on Build GRCh38
rs549161916 Chr.16: 88807499-88807499 on Build GRCh38
rs758810157 Chr.16: 88807441-88807441 on Build GRCh38
rs369852524 Chr.16: 88807509-88807509 on Build GRCh38
rs528610361 Chr.16: 88807383-88807383 on Build GRCh38
rs142465113 Chr.16: 88807394-88807394 on Build GRCh38
rs753287084 Chr.16: 88807498-88807498 on Build GRCh38
rs745962041 Chr.16: 88807468-88807468 on Build GRCh38
rs775339367 Chr.16: 88807375-88807375 on Build GRCh38
rs200652608 Chr.16: 88807407-88807407 on Build GRCh38
rs773348518 Chr.16: 88807504-88807505 on Build GRCh38
rs938840995 Chr.16: 88807388-88807388 on Build GRCh38

View Full Product Details