Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74591515-74591768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
UniGene
Hs.647642
Gene Chromosome Location
Chr.15: 74541177-74598131 on Build GRCh38
Transcripts
NM_001307939.1 NM_006465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370256 c.1232G>A p.S411N Chr.15: 74591626-74591626 on Build GRCh38
3731166 c.1250G>A p.R417Q Chr.15: 74591644-74591644 on Build GRCh38
3983041 c.1280G>A p.R427Q Chr.15: 74591674-74591674 on Build GRCh38
5798661 c.1317G>A p.S439S Chr.15: 74591711-74591711 on Build GRCh38
4623177 c.1264G>T p.E422* Chr.15: 74591658-74591658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775167120 Chr.15: 74591638-74591638 on Build GRCh38
rs758898092 Chr.15: 74591620-74591620 on Build GRCh38
rs569459248 Chr.15: 74591612-74591612 on Build GRCh38
rs202228664 Chr.15: 74591644-74591644 on Build GRCh38
rs772075921 Chr.15: 74591649-74591649 on Build GRCh38
rs746101449 Chr.15: 74591547-74591547 on Build GRCh38
rs780681529 Chr.15: 74591623-74591623 on Build GRCh38
rs754135318 Chr.15: 74591664-74591664 on Build GRCh38
rs752995560 Chr.15: 74591668-74591668 on Build GRCh38
rs140393498 Chr.15: 74591706-74591706 on Build GRCh38
rs555824373 Chr.15: 74591665-74591665 on Build GRCh38
rs780212786 Chr.15: 74591673-74591673 on Build GRCh38
rs994915879 Chr.15: 74591683-74591683 on Build GRCh38
rs778206808 Chr.15: 74591678-74591678 on Build GRCh38
rs146742821 Chr.15: 74591674-74591674 on Build GRCh38
rs139972092 Chr.15: 74591652-74591652 on Build GRCh38
rs749934899 Chr.15: 74591697-74591697 on Build GRCh38
rs767796273 Chr.15: 74591592-74591592 on Build GRCh38
rs994597946 Chr.15: 74591708-74591708 on Build GRCh38
rs113883275 Chr.15: 74591734-74591734 on Build GRCh38
rs756306380 Chr.15: 74591543-74591543 on Build GRCh38
rs190404425 Chr.15: 74591651-74591651 on Build GRCh38
rs534180198 Chr.15: 74591663-74591663 on Build GRCh38
rs768356045 Chr.15: 74591711-74591711 on Build GRCh38
rs772164167 Chr.15: 74591739-74591739 on Build GRCh38
rs755413625 Chr.15: 74591599-74591599 on Build GRCh38
rs745323100 Chr.15: 74591624-74591624 on Build GRCh38
rs1025638822 Chr.15: 74591724-74591724 on Build GRCh38
rs754188986 Chr.15: 74591571-74591571 on Build GRCh38
rs372219475 Chr.15: 74591643-74591643 on Build GRCh38
rs761845082 Chr.15: 74591648-74591648 on Build GRCh38
rs760504029 Chr.15: 74591747-74591747 on Build GRCh38
rs754338167 Chr.15: 74591647-74591647 on Build GRCh38
rs755691793 Chr.15: 74591704-74591704 on Build GRCh38
rs779409973 Chr.15: 74591705-74591705 on Build GRCh38
rs952769956 Chr.15: 74591725-74591725 on Build GRCh38
rs748708053 Chr.15: 74591613-74591613 on Build GRCh38
rs769352414 Chr.15: 74591633-74591633 on Build GRCh38
rs113614863 Chr.15: 74591733-74591733 on Build GRCh38
rs765661355 Chr.15: 74591667-74591667 on Build GRCh38
rs946077821 Chr.15: 74591568-74591568 on Build GRCh38
rs778862790 Chr.15: 74591723-74591723 on Build GRCh38

View Full Product Details