Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74864077-74864318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4862136 c.830G>T p.R277L Chr.17: 74864188-74864188 on Build GRCh38
6663007 c.902G>A p.R301H Chr.17: 74864266-74864266 on Build GRCh38
311159 c.947G>A p.S316N Chr.17: 74864221-74864221 on Build GRCh38
4783184 c.832C>T p.R278W Chr.17: 74864186-74864186 on Build GRCh38
3730024 c.900delC p.F301fs*20 Chr.17: 74864118-74864118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762141278 Chr.17: 74864203-74864203 on Build GRCh38
rs770841208 Chr.17: 74864098-74864098 on Build GRCh38
rs372052799 Chr.17: 74864266-74864266 on Build GRCh38
rs528032016 Chr.17: 74864277-74864277 on Build GRCh38
rs34118765 Chr.17: 74864249-74864249 on Build GRCh38
rs149273007 Chr.17: 74864174-74864174 on Build GRCh38
rs780646346 Chr.17: 74864166-74864166 on Build GRCh38
rs34495658 Chr.17: 74864140-74864140 on Build GRCh38
rs201513638 Chr.17: 74864156-74864156 on Build GRCh38
rs755725171 Chr.17: 74864248-74864248 on Build GRCh38
rs752410856 Chr.17: 74864170-74864170 on Build GRCh38
rs373171895 Chr.17: 74864267-74864267 on Build GRCh38
rs768923930 Chr.17: 74864194-74864194 on Build GRCh38
rs765580511 Chr.17: 74864136-74864136 on Build GRCh38
rs770805584 Chr.17: 74864185-74864185 on Build GRCh38
rs375205803 Chr.17: 74864205-74864205 on Build GRCh38
rs1048460112 Chr.17: 74864280-74864280 on Build GRCh38
rs755746794 Chr.17: 74864171-74864171 on Build GRCh38
rs763376429 Chr.17: 74864139-74864139 on Build GRCh38
rs575768148 Chr.17: 74864172-74864172 on Build GRCh38
rs769934690 Chr.17: 74864281-74864281 on Build GRCh38
rs960529836 Chr.17: 74864157-74864157 on Build GRCh38
rs202160207 Chr.17: 74864119-74864119 on Build GRCh38
rs773484169 Chr.17: 74864207-74864207 on Build GRCh38
rs200740021 Chr.17: 74864122-74864122 on Build GRCh38
rs760030067 Chr.17: 74864225-74864225 on Build GRCh38
rs372851948 Chr.17: 74864257-74864257 on Build GRCh38
rs752143061 Chr.17: 74864234-74864234 on Build GRCh38
rs552432 Chr.17: 74864131-74864131 on Build GRCh38
rs369448948 Chr.17: 74864256-74864256 on Build GRCh38
rs776839972 Chr.17: 74864199-74864199 on Build GRCh38
rs201450571 Chr.17: 74864278-74864278 on Build GRCh38
rs767069457 Chr.17: 74864233-74864233 on Build GRCh38
rs201770669 Chr.17: 74864120-74864120 on Build GRCh38
rs750745676 Chr.17: 74864137-74864137 on Build GRCh38
rs377052914 Chr.17: 74864105-74864105 on Build GRCh38
rs377041334 Chr.17: 74864097-74864097 on Build GRCh38
rs752096124 Chr.17: 74864218-74864218 on Build GRCh38
rs369953619 Chr.17: 74864109-74864109 on Build GRCh38
rs773536128 Chr.17: 74864286-74864286 on Build GRCh38
rs751057552 Chr.17: 74864154-74864154 on Build GRCh38
rs144469542 Chr.17: 74864189-74864189 on Build GRCh38
rs778860222 Chr.17: 74864186-74864186 on Build GRCh38
rs760795897 Chr.17: 74864117-74864117 on Build GRCh38
rs754527895 Chr.17: 74864164-74864164 on Build GRCh38
rs1022260408 Chr.17: 74864273-74864273 on Build GRCh38
rs908191097 Chr.17: 74864145-74864145 on Build GRCh38
rs948669536 Chr.17: 74864224-74864224 on Build GRCh38

View Full Product Details