Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47162425-47162696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM2
Entrez Gene ID
805
Gene Name
calmodulin 2
Gene Aliases
CAMII, LQT15, PHKD, PHKD2, caM
UniGene
Hs.468442
Gene Chromosome Location
Chr.2: 47160082-47176936 on Build GRCh38
Transcripts
NM_001305624.1 NM_001305625.1 NM_001305626.1 NM_001743.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5791013 c.112A>C p.R38R Chr.2: 47162585-47162585 on Build GRCh38
272457 c.55C>T p.L19L Chr.2: 47162642-47162642 on Build GRCh38
3839606 c.39C>G p.F13L Chr.2: 47162658-47162658 on Build GRCh38
1021232 c.116C>A p.S39Y Chr.2: 47162581-47162581 on Build GRCh38
1021233 c.77G>A p.G26E Chr.2: 47162620-47162620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781666473 Chr.2: 47162619-47162619 on Build GRCh38
rs754538204 Chr.2: 47162640-47162640 on Build GRCh38
rs116054300 Chr.2: 47162453-47162453 on Build GRCh38
rs1003389041 Chr.2: 47162452-47162452 on Build GRCh38
rs773024711 Chr.2: 47162488-47162488 on Build GRCh38
rs994243072 Chr.2: 47162465-47162465 on Build GRCh38
rs11551466 Chr.2: 47162580-47162580 on Build GRCh38
rs112786165 Chr.2: 47162626-47162626 on Build GRCh38
rs548280442 Chr.2: 47162512-47162512 on Build GRCh38
rs11551432 Chr.2: 47162586-47162586 on Build GRCh38
rs747792544 Chr.2: 47162665-47162665 on Build GRCh38
rs774005966 Chr.2: 47162477-47162477 on Build GRCh38
rs879026844 Chr.2: 47162511-47162511 on Build GRCh38
rs895938359 Chr.2: 47162462-47162462 on Build GRCh38
rs778389067 Chr.2: 47162644-47162644 on Build GRCh38
rs753038703 Chr.2: 47162547-47162547 on Build GRCh38
rs11551457 Chr.2: 47162629-47162629 on Build GRCh38
rs11551463 Chr.2: 47162550-47162550 on Build GRCh38
rs11551456 Chr.2: 47162615-47162615 on Build GRCh38
rs11551458 Chr.2: 47162582-47162582 on Build GRCh38
rs768087618 Chr.2: 47162468-47162468 on Build GRCh38
rs934381253 Chr.2: 47162502-47162502 on Build GRCh38
rs984097734 Chr.2: 47162479-47162479 on Build GRCh38
rs765398786 Chr.2: 47162538-47162538 on Build GRCh38
rs751977184 Chr.2: 47162592-47162592 on Build GRCh38
rs369374910 Chr.2: 47162574-47162574 on Build GRCh38
rs559299165 Chr.2: 47162466-47162466 on Build GRCh38
rs201394950 Chr.2: 47162478-47162478 on Build GRCh38
rs11551450 Chr.2: 47162548-47162548 on Build GRCh38
rs879199109 Chr.2: 47162513-47162513 on Build GRCh38
rs11551442 Chr.2: 47162572-47162572 on Build GRCh38
rs771731969 Chr.2: 47162668-47162668 on Build GRCh38
rs749244048 Chr.2: 47162496-47162497 on Build GRCh38
rs879070172 Chr.2: 47162514-47162514 on Build GRCh38
rs757523692 Chr.2: 47162593-47162593 on Build GRCh38
rs760575571 Chr.2: 47162494-47162494 on Build GRCh38
rs976949001 Chr.2: 47162636-47162636 on Build GRCh38
rs766437970 Chr.2: 47162504-47162504 on Build GRCh38
rs376209800 Chr.2: 47162507-47162507 on Build GRCh38

View Full Product Details