Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18742711-18742946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC1
Entrez Gene ID
23373
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
MECT1, TORC-1, TORC1, WAMTP1
UniGene
Hs.371096
Gene Chromosome Location
Chr.19: 18683615-18782333 on Build GRCh38
Transcripts
NM_001098482.1 NM_015321.2 XM_005259833.3 XM_005259834.1 XM_005259835.3 XM_005259836.3 XM_006722710.3 XM_011527842.2 XM_017026536.1 XM_017026537.1 XM_017026538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776035 c.127-7C>T p.? Chr.19: 18742903-18742903 on Build GRCh38
5776036 c.127-7C>T p.? Chr.19: 18742903-18742903 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942198893 Chr.19: 18742749-18742749 on Build GRCh38
rs374969669 Chr.19: 18742906-18742906 on Build GRCh38
rs748463661 Chr.19: 18742912-18742912 on Build GRCh38
rs140868731 Chr.19: 18742786-18742786 on Build GRCh38
rs763996458 Chr.19: 18742898-18742899 on Build GRCh38
rs200456495 Chr.19: 18742909-18742909 on Build GRCh38
rs1054851049 Chr.19: 18742753-18742753 on Build GRCh38
rs769035575 Chr.19: 18742908-18742908 on Build GRCh38
rs373049904 Chr.19: 18742903-18742903 on Build GRCh38
rs745554513 Chr.19: 18742885-18742885 on Build GRCh38
rs761016091 Chr.19: 18742774-18742774 on Build GRCh38
rs369866685 Chr.19: 18742884-18742884 on Build GRCh38
rs569938199 Chr.19: 18742905-18742905 on Build GRCh38
rs556685271 Chr.19: 18742770-18742770 on Build GRCh38
rs978281621 Chr.19: 18742769-18742769 on Build GRCh38
rs536789101 Chr.19: 18742757-18742757 on Build GRCh38
rs991041858 Chr.19: 18742815-18742815 on Build GRCh38
rs117770608 Chr.19: 18742927-18742927 on Build GRCh38
rs2075017 Chr.19: 18742743-18742743 on Build GRCh38
rs375809198 Chr.19: 18742869-18742869 on Build GRCh38
rs567628414 Chr.19: 18742744-18742744 on Build GRCh38
rs902207156 Chr.19: 18742857-18742857 on Build GRCh38
rs757058905 Chr.19: 18742876-18742876 on Build GRCh38

View Full Product Details