Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42964049-42964310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARSD1
Entrez Gene ID
80755
Gene Name
alanyl-tRNA synthetase domain containing 1
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42964498 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Gene Symbol
PTGES3L-AARSD1
Entrez Gene ID
100885850
Gene Name
PTGES3L-AARSD1 readthrough
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42980528 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5547777 c.423_426delTGAA p.E142fs*9 Chr.17: 42964190-42964193 on Build GRCh38
6018985 c.684C>T p.G228G Chr.17: 42964115-42964115 on Build GRCh38
5520249 c.475G>T p.E159* Chr.17: 42964141-42964141 on Build GRCh38
4642120 c.592G>A p.A198T Chr.17: 42964207-42964207 on Build GRCh38
5547776 c.606_609delTGAA p.E203fs*9 Chr.17: 42964190-42964193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974106168 Chr.17: 42964286-42964286 on Build GRCh38
rs779156060 Chr.17: 42964248-42964248 on Build GRCh38
rs909093806 Chr.17: 42964269-42964269 on Build GRCh38
rs373593609 Chr.17: 42964159-42964159 on Build GRCh38
rs749922163 Chr.17: 42964108-42964108 on Build GRCh38
rs753454107 Chr.17: 42964123-42964123 on Build GRCh38
rs690941 Chr.17: 42964217-42964217 on Build GRCh38
rs112590285 Chr.17: 42964280-42964280 on Build GRCh38
rs748750078 Chr.17: 42964133-42964134 on Build GRCh38
rs771976276 Chr.17: 42964169-42964169 on Build GRCh38
rs11278824 Chr.17: 42964275-42964280 on Build GRCh38
rs201921457 Chr.17: 42964207-42964207 on Build GRCh38
rs766025336 Chr.17: 42964114-42964114 on Build GRCh38
rs956495855 Chr.17: 42964239-42964239 on Build GRCh38
rs926583409 Chr.17: 42964193-42964193 on Build GRCh38
rs762765338 Chr.17: 42964076-42964076 on Build GRCh38
rs761747372 Chr.17: 42964083-42964083 on Build GRCh38
rs775611857 Chr.17: 42964075-42964075 on Build GRCh38
rs757731904 Chr.17: 42964208-42964208 on Build GRCh38
rs149954874 Chr.17: 42964276-42964281 on Build GRCh38
rs778004676 Chr.17: 42964140-42964140 on Build GRCh38
rs769792797 Chr.17: 42964158-42964158 on Build GRCh38
rs774071847 Chr.17: 42964166-42964167 on Build GRCh38
rs751855766 Chr.17: 42964258-42964258 on Build GRCh38
rs776808470 Chr.17: 42964278-42964282 on Build GRCh38
rs755146416 Chr.17: 42964244-42964244 on Build GRCh38
rs752036999 Chr.17: 42964149-42964149 on Build GRCh38
rs983953388 Chr.17: 42964105-42964105 on Build GRCh38
rs151255124 Chr.17: 42964153-42964153 on Build GRCh38
rs1053580119 Chr.17: 42964226-42964226 on Build GRCh38
rs548746113 Chr.17: 42964138-42964138 on Build GRCh38
rs773074132 Chr.17: 42964263-42964263 on Build GRCh38
rs200949352 Chr.17: 42964206-42964206 on Build GRCh38
rs780005682 Chr.17: 42964234-42964234 on Build GRCh38
rs747908457 Chr.17: 42964254-42964254 on Build GRCh38
rs762266502 Chr.17: 42964278-42964279 on Build GRCh38
rs761655197 Chr.17: 42964163-42964163 on Build GRCh38
rs746922327 Chr.17: 42964264-42964264 on Build GRCh38
rs773176232 Chr.17: 42964179-42964179 on Build GRCh38
rs1056668745 Chr.17: 42964134-42964134 on Build GRCh38
rs765818573 Chr.17: 42964196-42964196 on Build GRCh38
rs764709447 Chr.17: 42964277-42964280 on Build GRCh38
rs72039098 Chr.17: 42964286-42964291 on Build GRCh38
rs768094571 Chr.17: 42964214-42964214 on Build GRCh38
rs745909665 Chr.17: 42964155-42964155 on Build GRCh38
rs759107155 Chr.17: 42964204-42964204 on Build GRCh38
rs750653483 Chr.17: 42964282-42964283 on Build GRCh38
rs953210886 Chr.17: 42964245-42964245 on Build GRCh38
rs771886531 Chr.17: 42964261-42964261 on Build GRCh38
rs770551179 Chr.17: 42964143-42964151 on Build GRCh38
rs776279392 Chr.17: 42964197-42964197 on Build GRCh38
rs767512996 Chr.17: 42964100-42964100 on Build GRCh38
rs760140413 Chr.17: 42964185-42964185 on Build GRCh38
rs1053376928 Chr.17: 42964276-42964276 on Build GRCh38
rs146850790 Chr.17: 42964160-42964160 on Build GRCh38
rs796292768 Chr.17: 42964287-42964292 on Build GRCh38
rs770967729 Chr.17: 42964279-42964279 on Build GRCh38
rs1030615455 Chr.17: 42964230-42964230 on Build GRCh38
rs768361202 Chr.17: 42964078-42964078 on Build GRCh38
rs868263272 Chr.17: 42964287-42964287 on Build GRCh38
rs377670033 Chr.17: 42964240-42964240 on Build GRCh38
rs756164870 Chr.17: 42964233-42964233 on Build GRCh38
rs765372705 Chr.17: 42964281-42964285 on Build GRCh38
rs867214686 Chr.17: 42964288-42964288 on Build GRCh38
rs140103494 Chr.17: 42964110-42964110 on Build GRCh38

View Full Product Details